Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01718341_m1
          See other GM15688 GE Assays ›
          Gene Symbol:
          Gm15688
          Gene Name:
          predicted gene 15688
          Gene Aliases:
          EG433658, OTTMUSG00000025310
          Chromosome Location:
          Chr.3: 141457972 - 141465496 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01718341_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm15688
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK035802.1 - 1 - 2 729 86

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream pyruvate dehydrogenase E1 alpha 2 Pdha2 Pdhal -245.6kb Sense Chr. 3 - 141210004 - 141212355 on NCBI Build GRCm38
          Downstream unc-5 homolog C (C. elegans) Unc5c B130051O18Rik,
          Unc5h3,
          rcm
          231.0 Anti-sense Chr. 3 - 141465564 - 141834924 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2388991 AK035802.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 669512 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701447 AY916507.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701447 BC116952.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701447 NM_020502.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 604567 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 640809 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 17446 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151170 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2388991 AK035802.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151170 - -
          Whole Mouse Genome G4122A A_51_P376838 AK035802.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104163 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22584208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22584203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22584199 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22584193 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22584204 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22584182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22584179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22584189 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22584184 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22584214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22584191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22584187 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22584192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22584196 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22584209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22584211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22584206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22584215 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03002772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22584212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22584201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22584197 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 763699 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0