Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01718345_m1
          See other 1700030L20RIK GE Assays ›
          Gene Symbol:
          1700030L20Rik
          Gene Name:
          RIKEN cDNA 1700030L20 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 136435270 - 136449349 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01718345_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700030L20Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040592.1 4 - 5 698 74
          GenBank mRNA AK006559.1 - 4 - 5 699 74

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream B cell scaffold protein with ankyrin repeats 1 Bank1 A530094C12Rik,
          AI451642,
          AVIEF,
          BANK
          -109.1kb Sense Chr. 3 - 136053363 - 136326141 on NCBI Build GRCm38
          Downstream protein phosphatase 3,
          catalytic subunit,
          alpha isoform
          Ppp3ca 2900074D19Rik,
          CN,
          Caln,
          Calna,
          CnA
          232.4kb Anti-sense Chr. 3 - 136670066 - 136937727 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300003000.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2329938 AK006559.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 865935 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 18741 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558712 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 157772 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05086045 - -
          Whole Mouse Genome G4122A A_52_P890318 AK006559.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109337 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05086045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22557789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22557777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22557787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22557779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03028351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03028371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22557791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03072460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22557784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22557792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22557775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22557796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22557798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22557786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22557788 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300023694.mm.1 TC0300003000.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300023694.mm.1 NR_040592.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300023694.mm.1 NR_040592.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300023693.mm.1 TC0300003000.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300023693.mm.1 NR_040592.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300023693.mm.1 NR_040592.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300023696.mm.1 TC0300003000.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300023696.mm.1 NR_040592.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300023696.mm.1 NR_040592.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 350264 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0