Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01718645_s1
          See other AI428898 GE Assays ›
          Gene Symbol:
          AI428898
          Gene Name:
          expressed sequence AI428898
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01718645_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AI428898
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AI428898.1 - 1 103
          AK005805.1 - 1 761 103

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cyclin L2 Ccnl2 1700010A01Rik,
          1810019L15Rik,
          2010319M22Rik,
          AA409936,
          AW261738,
          Ania-6b,
          HCLA-150,
          Pcee,
          SB138
          Sense Chr. 4 - 155812084 - 155824543 on NCBI Build GRCm38
          Downstream aurora kinase A interacting protein 1 Aurkaip1 0610033H09Rik,
          Aip,
          Akip,
          MRP-S38
          18.9kb Sense Chr. 4 - 155831269 - 155833098 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2363883 AI428898.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2363883 AK005805.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174382 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174382 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174382 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10519208 - -
          Affymetrix HT MG-430 PM Array Plate 1459694_PM_at - -
          Mouse Genome 430 2.0 Array 1459694_at - -
          Mouse Genome 430 2.0 Array 1459694_PM_at - -
          Mouse Expression Set 430 1432195_s_at - -
          Mouse Expression Set 430 1453740_PM_a_at - -
          Mouse Expression Set 430 1454149_a_at - -
          Mouse Expression Set 430 1432195_PM_s_at - -
          Mouse Expression Set 430 1454149_PM_a_at - -
          Mouse Expression Set 430 1453740_a_at - -
          Mouse Expression Set 430 1459694_at - -
          Mouse Expression Set 430 1459694_PM_at - -
          Murine Genome U74v2 Set 102781_PM_at - -
          Murine Genome U74v2 Set 102781_at - -
          Murine Genome U74v2 Set 129253_PM_r_at - -
          Murine Genome U74v2 Set 129253_r_at - -
          Mouse Genome 430A 2.0 Array 1454149_a_at - -
          Mouse Genome 430A 2.0 Array 1453740_a_at - -
          Mouse Genome 430A 2.0 Array 1432195_s_at - -
          Mouse Genome 430A 2.0 Array 1432195_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1454149_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1453740_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1459694_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1459694_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 728801 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0