Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01718867_s1
          See other 4930431P19RIK GE Assays ›
          Gene Symbol:
          4930431P19Rik
          Gene Name:
          RIKEN cDNA 4930431P19 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01718867_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930431P19Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK031634.1 - 1 727 71
          AK050574.1 - 1 720 71

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream achaete-scute complex homolog 3 (Drosophila) Ascl3 Sgn1,
          bHLHa42
          -20.6kb Anti-sense Chr. 7 - 109727465 - 109731732 on NCBI Build GRCm38
          Downstream nuclear receptor interacting protein 3 Nrip3 A330103B05Rik,
          D7H11orf14,
          ICRFP703B1614Q5.2
          2.8kb Anti-sense Chr. 7 - 109758056 - 109781545 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P236528 AK031634.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P236528 AK031634.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 72555 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 11875 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119379 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152536 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P236528 AK031634.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P236528 AK031634.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152536 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119379 - -
          Whole Mouse Genome G4122A A_52_P236528 AK031634.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P236528 AK031634.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P147733 AK015280.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124867 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152536 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119379 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1429885_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1446949_PM_at - -
          Mouse Genome 430 2.0 Array 1429885_at - -
          Mouse Genome 430 2.0 Array 1446949_at - -
          Mouse Genome 430 2.0 Array 1446949_PM_at - -
          Mouse Genome 430 2.0 Array 1429885_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73886_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73886_at - -
          Mouse Expression Set 430 1446949_at - -
          Mouse Expression Set 430 1429885_at - -
          Mouse Expression Set 430 1446949_PM_at - -
          Mouse Expression Set 430 1429885_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429885_at - -
          Affymetrix HT Mouse Genome 430B Array 1446949_at - -
          Affymetrix HT Mouse Genome 430B Array 1446949_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429885_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 841320 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline