Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01718881_m1
          See other 4933406I18RIK GE Assays ›
          Gene Symbol:
          4933406I18Rik
          Gene Name:
          RIKEN cDNA 4933406I18 gene
          Gene Aliases:
          4933403D24Rik
          Chromosome Location:
          Chr.7: 114315479 - 114415161 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01718881_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4933406I18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029437.1 3 - 4 599 99
          GenBank mRNA AK016693.1 - 3 - 4 605 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004138.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170519 NR_029437.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 689795 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 830360 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 13525 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 457262 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765244 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 86754 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 182579 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 195039 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 248255 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 865303 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233053 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 924864 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 845399 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 673482 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 449793 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 718682 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 406763 AK083102.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 406763 AK083121.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 406763 AK158355.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 406763 NM_175650.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 962895 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930968 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 180155 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 869263 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 494492 AK038563.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 494492 AK136304.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 494492 AK154058.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 494492 NM_177622.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 90643 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 965208 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 863104 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 366400 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 117134 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 806898 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 899270 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 339358 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 474718 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 253379 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 549869 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 119250 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 462466 AK079788.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 447289 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 721109 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 99426 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 248640 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 576 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 330239 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 88393 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 347905 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 53464 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 71205 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 46308 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 160505 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64848 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64209 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 19425 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 134143 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86092 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P172310 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170519 NR_029437.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06330087 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01609027 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06329947 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P172310 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06330041 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06330056 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01609051 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06330073 - -
          Whole Mouse Genome G4122A A_52_P351495 NR_029438.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P351495 AK016630.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126273 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06330073 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06329947 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06330041 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P172310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05956596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06217829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06619568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06237537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05954966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06609634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06418540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06637732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06300258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06480376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06164959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06300260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05976716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06334168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06579693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06166634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06271004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06510631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06072309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06146685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06204323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06146691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25435352 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700034296.mm.1 TC0700004138.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700034296.mm.1 NR_029437.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700034296.mm.1 NR_029437.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 333444 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102470102 - -
          Illumina mouse-6 v1.0 expression beadchip 102470102 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247887 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215925 - -
          mouse-6 v1.1 (Illumina) scl00034.1_63-S - -
          Illumina Sentrix mouse-6.v1 bead array 102470102 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102470102 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0