Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01718955_g1
          See other IGHM GE Assays ›
          Gene Symbol:
          Ighm
          Gene Name:
          immunoglobulin heavy constant mu
          Gene Aliases:
          AI326478, Igh-6, Igh-M, Igh6, Igm, muH
          Chromosome Location:
          Chr.12: 113418826 - 113422730 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01718955_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ighm
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF052834.1 - 1331 78
          AI326478.1 - 1 - 2 78
          AJ294737.1 - 3 - 4 971 78
          AK039722.1 - 999 78
          AK050599.1 - 995 78
          AK088666.1 - 1183 78
          BC018315.1 - 1412 78
          BC053409.1 - 1432 78
          BC096667.1 - 1402 78
          BC098504.1 - 1431 78
          BC099618.1 - 1379 78
          V00827.1 - 3 - 4 971 78
          X03690.1 - 3 - 4 971 78

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P485421 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158540 - -
          Whole Mouse Genome G4122A A_51_P485421 - -
          Whole Mouse Genome G4122A A_52_P311042 - -
          Whole Mouse Genome G4122A A_52_P358360 BG070287.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P358360 V00821.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P358360 BC098504.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P358360 BC018315.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P358360 AK088666.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P28756575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28756560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10316392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28756565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28756566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28756579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28756567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10316397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28756557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28756564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28756563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28756588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28756582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28756577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28756581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28756584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28756573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28756571 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10402892 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402957 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402896 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402950 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402891 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402958 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402895 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402889 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402893 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402897 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402901 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402894 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402949 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402948 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402890 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403055 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402956 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402898 - -
          Affymetrix HT MG-430 PM Array Plate 1427756_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1427351_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1426196_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1427870_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1442544_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1452535_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1425324_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1427869_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10403054 - -
          Affymetrix Mouse Gene 1.1 ST Array 10402864 - -
          Mouse Genome 430 2.0 Array 1427869_PM_at - -
          Mouse Genome 430 2.0 Array 1427329_a_at - -
          Mouse Genome 430 2.0 Array 1426199_x_at - -
          Mouse Genome 430 2.0 Array 1455530_PM_at - -
          Mouse Genome 430 2.0 Array 1442544_at - -
          Mouse Genome 430 2.0 Array 1426196_PM_at - -
          Mouse Genome 430 2.0 Array 1426196_at - -
          Mouse Genome 430 2.0 Array 1426199_PM_x_at - -
          Mouse Genome 430 2.0 Array 1427351_s_at - -
          Mouse Genome 430 2.0 Array 1442544_PM_at - -
          Mouse Genome 430 2.0 Array 1427351_PM_s_at - -
          Mouse Genome 430 2.0 Array 1452535_PM_at - -
          Mouse Genome 430 2.0 Array 1427869_at - -
          Mouse Genome 430 2.0 Array 1426198_at - -
          Mouse Genome 430 2.0 Array 1427329_PM_a_at - -
          Mouse Genome 430 2.0 Array 1452535_at - -
          Mouse Genome 430 2.0 Array 1455530_at - -
          Mouse Genome 430 2.0 Array 1451958_PM_at - -
          Mouse Genome 430 2.0 Array 1451958_at - -
          Mouse Genome 430 2.0 Array 1426198_PM_at - -
          Mouse Expression Set 430 1426196_at - -
          Mouse Expression Set 430 1451947_at - -
          Mouse Expression Set 430 1452538_PM_at - -
          Mouse Expression Set 430 1452535_PM_at - -
          Mouse Expression Set 430 1452538_at - -
          Mouse Expression Set 430 1451949_at - -
          Mouse Expression Set 430 1452535_at - -
          Mouse Expression Set 430 1427869_at - -
          Mouse Expression Set 430 1427869_PM_at - -
          Mouse Expression Set 430 1427351_s_at - -
          Mouse Expression Set 430 1427758_x_at - -
          Mouse Expression Set 430 1427758_PM_x_at - -
          Mouse Expression Set 430 1451947_PM_at - -
          Mouse Expression Set 430 1451949_PM_at - -
          Mouse Expression Set 430 1426196_PM_at - -
          Mouse Expression Set 430 1427351_PM_s_at - -
          Mouse Expression Set 430 1442544_PM_at - -
          Mouse Expression Set 430 1442544_at - -
          Murine Genome U74v2 Set 97575_PM_f_at - -
          Murine Genome U74v2 Set 102159_at - -
          Murine Genome U74v2 Set 92470_PM_f_at - -
          Murine Genome U74v2 Set 101745_f_at - -
          Murine Genome U74v2 Set 93584_at - -
          Murine Genome U74v2 Set 101346_PM_at - -
          Murine Genome U74v2 Set 93583_PM_s_at - -
          Murine Genome U74v2 Set 99421_at - -
          Murine Genome U74v2 Set 100361_f_at - -
          Murine Genome U74v2 Set 99420_PM_at - -
          Murine Genome U74v2 Set 100362_f_at - -
          Murine Genome U74v2 Set 99420_at - -
          Murine Genome U74v2 Set 93583_s_at - -
          Murine Genome U74v2 Set 100361_PM_f_at - -
          Murine Genome U74v2 Set 101749_PM_at - -
          Murine Genome U74v2 Set 93584_PM_at - -
          Murine Genome U74v2 Set 101346_at - -
          Murine Genome U74v2 Set 101749_at - -
          Murine Genome U74v2 Set 99421_PM_at - -
          Murine Genome U74v2 Set 100362_PM_f_at - -
          Murine Genome U74v2 Set 97577_PM_f_at - -
          Murine Genome U74v2 Set 92470_f_at - -
          Murine Genome U74v2 Set 102159_PM_at - -
          Murine Genome U74v2 Set 97577_f_at - -
          Murine Genome U74v2 Set 97575_f_at - -
          Murine Genome U74v2 Set 101745_PM_f_at - -
          Murine Genome U74v2 Set 133672_at - -
          Murine Genome U74v2 Set 133672_PM_at - -
          Mouse Genome 430A 2.0 Array 1451949_at - -
          Mouse Genome 430A 2.0 Array 1427351_s_at - -
          Mouse Genome 430A 2.0 Array 1427758_x_at - -
          Mouse Genome 430A 2.0 Array 1452535_PM_at - -
          Mouse Genome 430A 2.0 Array 1427758_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1451949_PM_at - -
          Mouse Genome 430A 2.0 Array 1452538_PM_at - -
          Mouse Genome 430A 2.0 Array 1452535_at - -
          Mouse Genome 430A 2.0 Array 1451947_at - -
          Mouse Genome 430A 2.0 Array 1426196_at - -
          Mouse Genome 430A 2.0 Array 1427869_at - -
          Mouse Genome 430A 2.0 Array 1452538_at - -
          Mouse Genome 430A 2.0 Array 1427869_PM_at - -
          Mouse Genome 430A 2.0 Array 1427351_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1451947_PM_at - -
          Mouse Genome 430A 2.0 Array 1426196_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427351_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1452535_at - -
          Affymetrix HT Mouse Genome 430A Array 1427869_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427351_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1426196_at - -
          Affymetrix HT Mouse Genome 430A Array 1426196_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427869_at - -
          Affymetrix HT Mouse Genome 430A Array 1452535_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1442544_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1442544_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc