Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01719658_m1
          See other C330011F03RIK GE Assays ›
          Gene Symbol:
          C330011F03Rik
          Gene Name:
          RIKEN cDNA C330011F03 gene
          Gene Aliases:
          C330011F03
          Chromosome Location:
          Chr.17: 51425209 - 51448269 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01719658_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C330011F03Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046166.1 4 - 5 1590 70
          GenBank mRNA AK049176.1 - 4 - 5 1590 70

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream TBC1 domain family,
          member 5
          Tbc1d5 1600014N05Rik -245.0kb Sense Chr. 17 - 50733124 - 51180198 on NCBI Build GRCm38
          Downstream special AT-rich sequence binding protein 1 Satb1 2610306G12Rik,
          AW413156
          301.7kb Sense Chr. 17 - 51736187 - 51833497 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002264.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC0900000023.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 498180 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 601231 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 743363 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 56851 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 774662 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 29568 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08801832 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08801832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233763 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13362887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13332602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13458653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13149112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13063300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31234003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13503304 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13339914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13578154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13577969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13277776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13362439 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233767 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13463609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13466561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31234001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31233781 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700021258.mm.1 TC1700002264.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700021258.mm.1 NR_046166.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021258.mm.1 NR_046166.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 530863 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105220102 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0