Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01720641_m1
          See other FOXO6OS GE Assays ›
          Gene Symbol:
          Foxo6os
          Gene Name:
          forkhead box O6, opposite strand
          Gene Aliases:
          4732418A04Rik
          Chromosome Location:
          Chr.4: 120291135 - 120303893 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01720641_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Foxo6os
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK028616.1 - 1 - 2 483 82
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB612793.1 - 1 - 2 483 Chr. 4 - 120291350 - 120302226 [+] on Build GRCm38 82

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream forkhead box O6 Foxo6 -4.1kb Anti-sense Chr. 4 - 120267078 - 120287261 on NCBI Build GRCm38
          Downstream sex comb on midleg homolog 1 Scmh1 AI315320,
          AI851618,
          Scml1,
          Scml3
          103.1kb Sense Chr. 4 - 120405281 - 120530199 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 359017 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 3159 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 926323 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 252853 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 378777 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 712036 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 187588 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 104412 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163845 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05378574 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163845 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127702 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23250576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23250533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03787315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23250540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03862821 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23250538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03348613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03887840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03701351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04003728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23250536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04044642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04044639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03701352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03245938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23250569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23250561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04044646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03702235 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23250532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03894748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03825136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23250559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03825135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23250566 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 823559 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0