Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01721270_m1
          See other 4930474H20RIK GE Assays ›
          Gene Symbol:
          4930474H20Rik
          Gene Name:
          RIKEN cDNA 4930474H20 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 90126891 - 90269760 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01721270_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930474H20Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045712.1 8 - 9 1357 145
          GenBank mRNA AK015575.1 - 8 - 9 1364 145

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream poly(A)-binding protein,
          cytoplasmic pseudogene
          Gm5088 EG328451,
          Pabpc4l
          -364.5kb Sense Chr. 14 - 89896224 - 89899447 on NCBI Build GRCm38
          Downstream protocadherin 9 Pcdh9 2.7mb Anti-sense Chr. 14 - 93013700 - 93890721 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400001137.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC1500000950.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2333797 AK015575.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914631 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 359953 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 388291 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 819202 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 938489 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 465324 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 13789 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 792393 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 466518 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 465535 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 20927 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 124897 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 749064 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 327550 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 85181 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 260659 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 622749 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930954 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 152775 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 53415 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 311798 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 66726 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 644945 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169517 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701278 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 808518 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 270505 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 485413 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 297308 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 278590 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 208619 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 969020 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 778252 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 645736 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 132934 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 940602 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 607164 AK049111.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 607164 XM_981774.2 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 607164 XM_986525.2 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 311481 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 491673 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 249122 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 167431 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 580903 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 466590 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 535108 NM_146775.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 869435 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 725214 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 271908 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 756001 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 503245 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 27290 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 38529 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 126229 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 125408 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 53078 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 16003 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 155784 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 174108 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 75069 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08195895 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08196066 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08195955 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08196050 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08196087 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08195962 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02909133 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08195919 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08195987 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08196015 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08195886 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08196031 - -
          Whole Mouse Genome G4122A A_52_P1035446 AK015575.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02909133 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08196087 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08195987 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08196031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29801189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29801174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11257465 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29801177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29801186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29801163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29801181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29801184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11680162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29801161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29801166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29801176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29801171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11617713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29801162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800780 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29801164 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29801185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29801165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29801180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29801167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29801178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29801170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29801173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29800966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29801183 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432058_PM_at - -
          Mouse Genome 430 2.0 Array 1432058_at - -
          Mouse Genome 430 2.0 Array 1432058_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1400009025.mm.1 TC1400001137.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400009025.mm.1 NR_045712.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009025.mm.1 NR_045712.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009026.mm.1 TC1400001137.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400009026.mm.1 NR_045712.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009026.mm.1 NR_045712.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009027.mm.1 TC1400001137.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400009027.mm.1 NR_045712.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009027.mm.1 NR_045712.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009028.mm.1 TC1400001137.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400009028.mm.1 NR_045712.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009028.mm.1 NR_045712.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009022.mm.1 TC1400001137.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400009022.mm.1 NR_045712.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009022.mm.1 NR_045712.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009023.mm.1 TC1400001137.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400009023.mm.1 NR_045712.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009023.mm.1 NR_045712.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009024.mm.1 TC1400001137.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400009024.mm.1 NR_045712.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009024.mm.1 NR_045712.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009021.mm.1 TC1400001137.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400009021.mm.1 NR_045712.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009021.mm.1 NR_045712.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009020.mm.1 TC1400001137.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400009020.mm.1 NR_045712.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009020.mm.1 NR_045712.1 Refseq
          Mouse Expression Set 430 1432058_PM_at - -
          Mouse Expression Set 430 1432058_at - -
          Affymetrix HT Mouse Genome 430B Array 1432058_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432058_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 823216 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0