Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01721443_m1
          See other GM20187 GE Assays ›
          Gene Symbol:
          Gm20187
          Gene Name:
          predicted gene, 20187
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 27090237 - 27114995 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01721443_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm20187
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045052.1 2 - 3 699 68
          GenBank mRNA AK084401.1 - 2 - 3 700 68

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 4933409F18 gene 4933409F18Rik 4930435E18Rik -255.3kb Sense Chr. 12 - 26814375 - 26834888 on NCBI Build GRCm38
          Downstream predicted gene 9866 Gm9866 EG636791,
          ENSMUSG00000052076
          43.9kb Sense Chr. 12 - 27140796 - 27160516 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001557.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 34342 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 681224 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 369776 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 275227 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 495357 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 362327 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 787957 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 111834 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159684 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159684 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02477140 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104744 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28305815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28305806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28305811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28305836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28305810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28305882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28305845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28305822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28305837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28305817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09923422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28305812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09923408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28305819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28305832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28305823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28305847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28305830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10169706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09824517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28305848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28305844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09923412 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200010779.mm.1 TC1200001557.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200010779.mm.1 NR_045052.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010779.mm.1 NR_045052.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 607023 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0