Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01721581_m1
          See other A730049N16RIK GE Assays ›
          Gene Symbol:
          A730049N16Rik
          Gene Name:
          RIKEN cDNA A730049N16 gene
          Gene Aliases:
          Gm548
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01721581_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A730049N16Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK043040.1 - 1 - 2 725 118

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA A930029G22 gene A930029G22Rik -33.5kb Sense Chr. 17 - 69356361 - 69440118 on NCBI Build GRCm38
          Downstream predicted gene,
          36594
          Gm36594 418.8kb Sense Chr. 17 - 69894630 - 69897331 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent ABI

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13567984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13092846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13112739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31324212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31324181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31324186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31324214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13460076 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13460061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31324187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31324220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31324185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31324209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13460065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13567977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31324217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13228052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31324219 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 538366 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix