Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01722231_g1
          See other 4930554I06RIK GE Assays ›
          Gene Symbol:
          4930554I06Rik
          Gene Name:
          RIKEN cDNA 4930554I06 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 21104710 - 21105538 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01722231_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930554I06Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK016120.1 - 1 - 2 43 56
          BY715723.1 - 1 - 2 43 56

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream transmembrane channel-like gene family 1 Tmc1 4933416G09Rik,
          Beethoven,
          Bth,
          CWEA1,
          dn
          -67.6kb Anti-sense Chr. 19 - 20783458 - 21037152 on NCBI Build GRCm38
          Downstream predicted gene 3775 Gm3775 15.6kb Anti-sense Chr. 19 - 21121152 - 21125705 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14341827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14247059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14263044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14444604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14503743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14368601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14503760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14352550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14368600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32004651 - -
          Array Name Probe IDs Targets Target Type
          Mouse Expression Set 430 1459487_PM_at - -
          Mouse Expression Set 430 1459487_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 308517 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline