Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01729473_gH
          See other ZFP950 GE Assays ›
          Gene Symbol:
          Zfp950
          Gene Name:
          zinc finger prtoein 950
          Gene Aliases:
          5830428H23Rik, AU016073, BC055757, Gm34518, Zfp826, mszf84
          Chromosome Location:
          Chr.19: 61051147 - 61313507 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01729473_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp950
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK045970.1 - 2 - 3 211 162
          AK164548.1 - 2 - 3 218 162
          BC055757.1 - 2 - 3 191 162

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2208300 AK029211.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 308 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 389951 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732463 AK003144.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732463 AK085114.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732463 AK134275.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732463 AK151062.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732463 AK153398.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732463 AK159605.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732463 AK170436.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732463 NM_010333.4 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 692179 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 284690 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 414168 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 140948 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 808821 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 492515 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 143696 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 828908 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 120175 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 538422 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 756236 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127316 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 144450 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P138472 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P135534 BC086681.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P135534 AK047931.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P135534 AK139999.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P135534 AK039536.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P135534 BC029127.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2208300 AK029211.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P279476 AK029211.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P207718 AK164548.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P207718 AK020020.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P207718 AI117149.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32229083 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10468886 - -
          Affymetrix Mouse Gene 1.0 ST Array 10468888 - -
          Affymetrix Mouse Gene 1.0 ST Array 10468883 - -
          Affymetrix Mouse Gene 1.0 ST Array 10468882 - -
          Affymetrix Mouse Gene 1.0 ST Array 10468884 - -
          Affymetrix HT MG-430 PM Array Plate 1436072_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1428748_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1438465_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10468885 - -
          Affymetrix Mouse Gene 1.1 ST Array 10468881 - -
          Mouse Genome 430 2.0 Array 1436072_at - -
          Mouse Genome 430 2.0 Array 1428748_PM_at - -
          Mouse Genome 430 2.0 Array 1428748_at - -
          Mouse Genome 430 2.0 Array 1438465_at - -
          Mouse Genome 430 2.0 Array 1438465_PM_at - -
          Mouse Genome 430 2.0 Array 1436072_PM_at - -
          Mouse Expression Set 430 1438465_PM_at - -
          Mouse Expression Set 430 1438465_at - -
          Mouse Expression Set 430 1436072_at - -
          Mouse Expression Set 430 1428748_at - -
          Mouse Expression Set 430 1428748_PM_at - -
          Mouse Expression Set 430 1436072_PM_at - -
          Mouse Genome 430A 2.0 Array 1438465_PM_at - -
          Mouse Genome 430A 2.0 Array 1438465_at - -
          Affymetrix HT Mouse Genome 430A Array 1438465_at - -
          Affymetrix HT Mouse Genome 430A Array 1438465_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428748_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428748_at - -
          Affymetrix HT Mouse Genome 430B Array 1436072_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436072_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101410546 - -
          MouseWG-6 v2 Expression BeadChip 100360465 - -
          Illumina mouse-6 v1.0 expression beadchip 105360341 - -
          Illumina mouse-6 v1.0 expression beadchip 101410546 - -
          Illumina mouse-6 v1.0 expression beadchip 103290021 - -
          Illumina mouse-6 v1.0 expression beadchip 100360465 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258205 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222245 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256986 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1241044 - -
          Illumina Sentrix mouse-6.v1 bead array 105360341 - -
          Illumina Sentrix mouse-6.v1 bead array 100360465 - -
          Illumina Sentrix mouse-6.v1 bead array 103290021 - -
          Illumina Sentrix mouse-6.v1 bead array 101410546 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103290021 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101410546 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100360465 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105360341 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2557452 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1241044 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1222245 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1258205 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline