Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01731378_g1
          See other PTGES3 GE Assays ›
          Gene Symbol:
          Ptges3
          Gene Name:
          prostaglandin E synthase 3 (cytosolic)
          Gene Aliases:
          5730442A20Rik, Ptges, Tebp, cPGES, p23, sid3177
          Chromosome Location:
          Chr.10: 128058982 - 128077254 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01731378_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptges3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019766.4 NP_062740.1 1 - 2 306 175
          GenBank mRNA AK008805.1 - 1 - 2 337 175
          AK010636.1 - 1 - 2 308 175
          AK075932.1 - 1 - 2 171 175
          AK075987.1 - 1 - 2 217 175
          AK077538.1 - 1 - 2 307 175
          AK167443.1 - 1 - 2 282 175
          AK168073.1 - 1 - 2 217 175
          AK168210.1 - 1 - 2 217 175
          AK168721.1 - 1 - 2 156 175
          AY281130.1 - 1 - 2 333 175
          BC003708.1 - 1 - 2 199 3489418 175
          BC085264.1 - 1 - 2 203 6392001 175

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000001581.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK008805.1, AK010636.1, AK075932.1, AK075987.1, AK077538.1, AK167443.1, AK168073.1, AK168210.1, AK168721.1, AY281130.1, BC003708.1, BC085264.1, NM_019766.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ptges3

          Gene Ontology Categories:

          Function(s) Process(es)

          prostaglandin biosynthetic process
          glycogen biosynthetic process
          RNA-dependent DNA biosynthetic process
          lipid metabolic process
          fatty acid metabolic process
          fatty acid biosynthetic process
          prostaglandin metabolic process
          cell population proliferation
          positive regulation of gene expression
          sensory perception of pain
          glucocorticoid receptor signaling pathway
          skin development
          chaperone cofactor-dependent protein refolding
          lung saccule development
          negative regulation of cell death
          DNA biosynthetic process
          p53 binding
          telomerase activity
          isomerase activity
          enzyme binding
          prostaglandin-E synthase activity
          unfolded protein binding
          Hsp90 protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000011437.mm.1 TC1000001581.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000011437.mm.1 NM_019766.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011437.mm.1 NM_019766.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011448.mm.1 TC1000001581.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000011448.mm.1 NM_019766.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011448.mm.1 NM_019766.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011444.mm.1 TC1000001581.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000011444.mm.1 NM_019766.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011444.mm.1 NM_019766.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011447.mm.1 TC1000001581.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000011447.mm.1 NM_019766.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011447.mm.1 NM_019766.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011442.mm.1 TC1000001581.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000011442.mm.1 NM_019766.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011442.mm.1 NM_019766.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011438.mm.1 TC1000001581.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000011438.mm.1 NM_019766.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011438.mm.1 NM_019766.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011440.mm.1 TC1000001581.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000011440.mm.1 NM_019766.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011440.mm.1 NM_019766.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011445.mm.1 TC1000001581.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000011445.mm.1 NM_019766.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011445.mm.1 NM_019766.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011449.mm.1 TC1000001581.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000011449.mm.1 NM_019766.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011449.mm.1 NM_019766.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011441.mm.1 TC1000001581.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000011441.mm.1 NM_019766.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011441.mm.1 NM_019766.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 907427 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0