Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900023015.mm.1 |
TC0900002780.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023015.mm.1 |
NM_001198933.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023015.mm.1 |
NM_001198933.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023015.mm.1 |
NM_008615.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023015.mm.1 |
NM_008615.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023013.mm.1 |
TC0900002780.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023013.mm.1 |
NM_001198933.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023013.mm.1 |
NM_001198933.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023013.mm.1 |
NM_008615.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023013.mm.1 |
NM_008615.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023012.mm.1 |
TC0900002780.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023012.mm.1 |
NM_001198933.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023012.mm.1 |
NM_001198933.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023012.mm.1 |
NM_008615.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023012.mm.1 |
NM_008615.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023016.mm.1 |
TC0900002780.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023016.mm.1 |
NM_001198933.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023016.mm.1 |
NM_001198933.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023016.mm.1 |
NM_008615.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023016.mm.1 |
NM_008615.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023017.mm.1 |
TC0900002780.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023017.mm.1 |
NM_001198933.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023017.mm.1 |
NM_001198933.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023017.mm.1 |
NM_008615.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023017.mm.1 |
NM_008615.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023014.mm.1 |
TC0900002780.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023014.mm.1 |
NM_001198933.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023014.mm.1 |
NM_001198933.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023014.mm.1 |
NM_008615.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023014.mm.1 |
NM_008615.2 |
Refseq |