Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01960904_s1
          See other 1700073E17RIK GE Assays ›
          Gene Symbol:
          1700073E17Rik
          Gene Name:
          ribosomal protein L7 pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 145387625 - 145392224 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01960904_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700073E17Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003625.1 1 4384 93
          GenBank mRNA BC049631.1 - 2 856 93
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB679532.1 - 1 197 Chr. 6 - 145391812 - 145392230 [+] on Build GRCm38 93

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 15706 Gm15706 OTTMUSG00000025463 -139.6kb Sense Chr. 6 - 145250552 - 145251856 on NCBI Build GRCm38
          Downstream predicted gene,
          38475
          Gm38475 61.1kb Anti-sense Chr. 6 - 145453345 - 145614319 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001718.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P352563 BC049631.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P352563 NR_003625.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P352563 BC049631.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P352563 NR_003625.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 240401 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 828115 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 659688 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P352563 BC049631.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P352563 NR_003625.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P352563 BC049631.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P352563 NR_003625.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P424021 BC049631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P424021 NR_003625.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P424021 AK006943.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P352563 BC049631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P352563 NR_003625.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P352563 BC049631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P352563 NR_003625.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05833107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05308630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05027419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05573539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05062133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05417608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05833110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922684 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10542726 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542725 - -
          Affymetrix HT MG-430 PM Array Plate 1420257_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1420258_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1447611_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10542724 - -
          Mouse Genome 430 2.0 Array 1420258_at - -
          Mouse Genome 430 2.0 Array 1420258_PM_at - -
          Mouse Genome 430 2.0 Array 1420257_at - -
          Mouse Genome 430 2.0 Array 1447611_at - -
          Mouse Genome 430 2.0 Array 1420257_PM_at - -
          Mouse Genome 430 2.0 Array 1447611_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 381827_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 381827_at - -
          Affymetrix Mouse Clariom S Array PSR0600013655.mm.1 TC0600001718.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600013655.mm.1 NR_003625.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600013659.mm.1 TC0600001718.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600013659.mm.1 NR_003625.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600013652.mm.1 TC0600001718.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600013652.mm.1 NR_003625.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600013657.mm.1 TC0600001718.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600013657.mm.1 NR_003625.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600013653.mm.1 TC0600001718.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600013653.mm.1 NR_003625.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600013658.mm.1 TC0600001718.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600013658.mm.1 NR_003625.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600013654.mm.1 TC0600001718.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600013654.mm.1 NR_003625.1 Refseq
          Mouse Expression Set 430 1420257_at - -
          Mouse Expression Set 430 1420258_PM_at - -
          Mouse Expression Set 430 1420258_at - -
          Mouse Expression Set 430 1420257_PM_at - -
          Mouse Expression Set 430 1447611_at - -
          Mouse Expression Set 430 1447611_PM_at - -
          Murine Genome U74v2 Set 161226_f_at - -
          Murine Genome U74v2 Set 161226_PM_f_at - -
          Murine Genome U74v2 Set 169686_PM_r_at - -
          Murine Genome U74v2 Set 169686_r_at - -
          Mouse Genome 430A 2.0 Array 1420258_at - -
          Mouse Genome 430A 2.0 Array 1420257_PM_at - -
          Mouse Genome 430A 2.0 Array 1420258_PM_at - -
          Mouse Genome 430A 2.0 Array 1420257_at - -
          Affymetrix HT Mouse Genome 430A Array 1420258_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420258_at - -
          Affymetrix HT Mouse Genome 430A Array 1420257_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420257_at - -
          Affymetrix HT Mouse Genome 430B Array 1447611_at - -
          Affymetrix HT Mouse Genome 430B Array 1447611_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 520015 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2790902 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2790902 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0