Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01966262_s1
          See other AA543186 GE Assays ›
          Gene Symbol:
          AA543186
          Gene Name:
          expressed sequence AA543186
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 25327450 - 25332571 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01966262_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AA543186
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027448.1 2 934 130
          GenBank mRNA AK146156.1 - 2 935 130

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream glutamate receptor,
          ionotropic,
          NMDA1 (zeta 1)
          Grin1 GluN1,
          GluRdelta1,
          GluRzeta1,
          M100174,
          NMD-R1,
          NMDAR1,
          NR1,
          Nmdar,
          Rgsc174
          -8.3kb Sense Chr. 2 - 25291177 - 25319198 on NCBI Build GRCm38
          Downstream mannosidase,
          alpha,
          class 1B,
          member 1
          Man1b1 E430019H13Rik,
          ERMan1,
          Gm108,
          MANA-ER
          4.0kb Anti-sense Chr. 2 - 25332743 - 25352213 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003144.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 162588 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 823202 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 274217 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 527953 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 9867 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126158 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00324831 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126158 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100630 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126158 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200025930.mm.1 TC0200003144.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200025930.mm.1 NR_027448.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025930.mm.1 NR_027448.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025929.mm.1 TC0200003144.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200025929.mm.1 NR_027448.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025929.mm.1 NR_027448.1 Refseq
          Murine Genome U74v2 Set 136538_at - -
          Murine Genome U74v2 Set 136538_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 891048 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0