Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01971449_s1
          See other A230072C01RIK GE Assays ›
          Gene Symbol:
          A230072C01Rik
          Gene Name:
          RIKEN cDNA A230072C01 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 20961676 - 20987349 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01971449_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A230072C01Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027445.1 6 676 143
          NR_027446.1 4 922 143
          GenBank mRNA AK029359.1 - 6 669 143
          AK038883.1 - 4 923 143
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AV248189.2 - 1 162 Chr. X - 20986087 - 20986723 [+] on Build GRCm38 143
          GenBankmRNA BB178169.2 - 1 139 Chr. X - 20986138 - 20986737 [+] on Build GRCm38 143
          GenBankmRNA DT931353.1 - 1 153 Chr. X - 20986095 - 20986701 [+] on Build GRCm38 143

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ubiquitously expressed transcript Uxt 0910002B17Rik -24.0kb Anti-sense Chr. X - 20951665 - 20962035 on NCBI Build GRCm38
          Downstream zinc finger protein 182 Zfp182 6330583I20Rik,
          Znf182
          38.8kb Anti-sense Chr. X - 21026184 - 21062038 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00000221.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P673917 NR_027446.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P673917 NR_027445.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P673917 AK029359.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P673917 AK038883.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P673917 NR_027446.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P673917 NR_027445.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P673917 AK029359.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P673917 AK038883.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 957840 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 73468 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 600800 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 818963 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 846733 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 747638 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 856884 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 333151 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 162471 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 24154 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 49807 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P673917 NR_027446.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P673917 NR_027445.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P673917 AK029359.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P673917 AK038883.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P673917 NR_027446.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P673917 NR_027445.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P673917 AK029359.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P673917 AK038883.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09250539 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09250549 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09250557 - -
          Whole Mouse Genome G4122A A_52_P118771 - -
          Whole Mouse Genome G4122A A_52_P673917 NR_027446.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P673917 NR_027445.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P673917 AK029359.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P673917 AK038883.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P673917 NR_027446.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P673917 NR_027445.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P673917 AK029359.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P673917 AK038883.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138437 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09250539 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09250557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14612792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290930 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1445380_PM_at - -
          Mouse Genome 430 2.0 Array 1445380_at - -
          Mouse Genome 430 2.0 Array 1445380_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 320742_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 320742_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00001552.mm.1 TC0X00000221.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00001552.mm.1 NR_027445.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001552.mm.1 NR_027445.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001552.mm.1 NR_027446.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001552.mm.1 NR_027446.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001555.mm.1 TC0X00000221.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00001555.mm.1 NR_027445.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001555.mm.1 NR_027445.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001555.mm.1 NR_027446.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001555.mm.1 NR_027446.1 Refseq
          Mouse Expression Set 430 1445380_PM_at - -
          Mouse Expression Set 430 1445380_at - -
          Affymetrix HT Mouse Genome 430B Array 1445380_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1445380_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 507013 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105290731 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0