Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01972781_s1
          See other 4930432J09RIK GE Assays ›
          Gene Symbol:
          4930432J09Rik
          Gene Name:
          RIKEN cDNA 4930432J09 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 103977628 - 104027142 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01972781_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930432J09Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045953.1 3 994 118
          GenBank mRNA AK015289.1 - 3 948 118
          AK052661.1 - 3 995 118

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream endothelin receptor type B Ednrb ET-B,
          ET-BR,
          ETR-b,
          ETb,
          Sox10m1
          -133.2kb Sense Chr. 14 - 103814615 - 103844476 on NCBI Build GRCm38
          Downstream RIKEN cDNA D130079A08 gene D130079A08Rik 305.6kb Sense Chr. 14 - 104285845 - 104460120 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400002643.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC1500002298.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2402258 AK052661.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 719085 AF060244.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 719085 AF060245.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 719085 AF060246.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 719085 AK029313.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 719085 NM_011743.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626510 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 40583 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 291881 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 863417 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 272513 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 301695 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897806 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 35547 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 757426 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 237422 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 353763 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 748713 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 568452 AK030010.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 568452 BC132479.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 568452 EF199807.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 568452 NM_173023.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 940625 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584693 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 49973 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08223710 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02930303 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08223735 - -
          Whole Mouse Genome G4122A A_52_P308593 AK052661.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P308593 AK015289.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11160635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11204805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11626948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11703044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11626931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11812040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11562444 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869098 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11703046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11451250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11656199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11648303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11206618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11667393 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11560321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11656198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11626927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29869110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29868924 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432324_PM_at - -
          Mouse Genome 430 2.0 Array 1432324_PM_at - -
          Mouse Genome 430 2.0 Array 1432324_at - -
          Affymetrix Mouse Clariom S Array PSR1400019792.mm.1 TC1400002643.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400019792.mm.1 NR_045953.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400019792.mm.1 NR_045953.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400019793.mm.1 TC1400002643.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400019793.mm.1 NR_045953.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400019793.mm.1 NR_045953.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400019795.mm.1 TC1400002643.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400019795.mm.1 NR_045953.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400019795.mm.1 NR_045953.1 Refseq
          Mouse Expression Set 430 1432324_PM_at - -
          Mouse Expression Set 430 1432324_at - -
          Affymetrix HT Mouse Genome 430B Array 1432324_at - -
          Affymetrix HT Mouse Genome 430B Array 1432324_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 678023 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106760400 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0