Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01973796_s1
          See other RBM12B1 GE Assays ›
          Gene Symbol:
          Rbm12b1
          Gene Name:
          RNA binding motif protein 12 B1
          Gene Aliases:
          3000004N20Rik, Rbm12b, Rbm12ba
          Chromosome Location:
          Chr.4: 12138957 - 12146745 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01973796_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rbm12b1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028226.2 NP_082502.2 3 2729 117
          GenBank mRNA AK013872.1 - 1 1824 117
          BC050844.1 - 3 2530 6513134 117
          BC054081.1 - 1 1462 117

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400000108.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rbm12b1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          nucleotide binding
          nucleic acid binding
          RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22733424 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P163173 BC054081.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P163173 BP767112.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P163173 BC031713.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P163173 BC050844.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P163173 AK013872.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P163173 BC054081.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P163173 BP767112.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P163173 BC031713.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P163173 BC050844.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P163173 AK013872.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 318382 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P163173 BC054081.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P163173 BP767112.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P163173 BC031713.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P163173 BC050844.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P163173 AK013872.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P163173 BC054081.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P163173 BP767112.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P163173 BC031713.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P163173 BC050844.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P163173 AK013872.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22733418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22733419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22733416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22733415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22733424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22733420 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22733412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22733423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22733414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22733417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22733422 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05157790 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05157779 - -
          Whole Mouse Genome G4122A A_52_P478289 BC137620.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P478289 BC030408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P478289 BC080741.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P478289 AK080943.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P478289 AK169895.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P354598 AK031339.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P163173 BC054081.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P163173 BP767112.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P163173 BC031713.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P163173 BC050844.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P163173 AK013872.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P163173 BC054081.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P163173 BP767112.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P163173 BC031713.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P163173 BC050844.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P163173 AK013872.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129998 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124233 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03866334 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03866332 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03632573 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22733424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22733418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22733489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22733414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22733395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22733393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22733485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22733423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22733396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22733536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22733416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22733422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22733415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22733487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22733409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22733486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22733420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22733411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22733416 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22733410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22733415 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22733486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03531115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22733489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22733447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22733423 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22733414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22733412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22733422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22733411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03489148 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22733396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22733420 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22733440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03555704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03866325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03257876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22733405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22733417 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03866361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22733393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22733395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22733407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22733485 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03751550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22733418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22733421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22733487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22733476 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10503366 - -
          Affymetrix Mouse Gene 1.0 ST Array 10503369 - -
          Affymetrix Mouse Gene 1.0 ST Array 10503364 - -
          Affymetrix Mouse Gene 1.0 ST Array 10503368 - -
          Affymetrix Mouse Gene 1.0 ST Array 10503365 - -
          Affymetrix Mouse Gene 1.0 ST Array 10503367 - -
          Affymetrix HT MG-430 PM Array Plate 1435158_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1453979_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10503363 - -
          Mouse Genome 430 2.0 Array 1435158_at - -
          Mouse Genome 430 2.0 Array 1435158_PM_at - -
          Mouse Genome 430 2.0 Array 1453979_PM_at - -
          Mouse Genome 430 2.0 Array 1453979_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72397_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72397_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400000743.mm.1 TC0400000108.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400000743.mm.1 NM_028226.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000743.mm.1 NM_028226.2 Refseq
          Mouse Expression Set 430 1453979_PM_at - -
          Mouse Expression Set 430 1453979_at - -
          Mouse Expression Set 430 1435158_at - -
          Mouse Expression Set 430 1435158_PM_at - -
          Murine Genome U74v2 Set 165652_PM_r_at - -
          Murine Genome U74v2 Set 165652_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1435158_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435158_at - -
          Affymetrix HT Mouse Genome 430B Array 1453979_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453979_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102120039 - -
          MouseWG-6 v2 Expression BeadChip 6380019 - -
          Illumina mouse-6 v1.0 expression beadchip 102120039 - -
          Illumina mouse-6 v1.0 expression beadchip 6380019 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006380019 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1240675 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218028 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1240675 - -
          mouse-6 v1.1 (Illumina) scl072397.3_51-S - -
          Illumina Sentrix mouse-6.v1 bead array 102120039 - -
          Illumina Sentrix mouse-6.v1 bead array 6380019 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl072397.3_51-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102120039 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6380019 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1240675 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1240675 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1218028 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0