Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01976234_s1
          See other 4933407G14RIK GE Assays ›
          Gene Symbol:
          4933407G14Rik
          Gene Name:
          RIKEN cDNA 4933407G14 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 75449510 - 75463104 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01976234_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4933407G14Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045841.1 1 829 143
          GenBank mRNA AK016719.1 - 1 830 143

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ureidopropionase,
          beta
          Upb1 AI195023 -7.8kb Sense Chr. 10 - 75406911 - 75441679 on NCBI Build GRCm38
          Downstream guanylyl cyclase domain containing 1 Gucd1 1110038D17Rik 56.4kb Anti-sense Chr. 10 - 75506814 - 75517967 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000715.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2200890 AK016719.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1138699 AK016719.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08628254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27256627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27256635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27256658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08377487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08339480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27256626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27256660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27256659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27256633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27256650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27256638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27256637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27256651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27256624 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1453937_PM_at - -
          Mouse Genome 430 2.0 Array 1453937_PM_at - -
          Mouse Genome 430 2.0 Array 1453937_at - -
          Affymetrix Mouse Clariom S Array PSR1000005082.mm.1 TC1000000715.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000005082.mm.1 NR_045841.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005082.mm.1 NR_045841.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005080.mm.1 TC1000000715.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000005080.mm.1 NR_045841.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005080.mm.1 NR_045841.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005081.mm.1 TC1000000715.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000005081.mm.1 NR_045841.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005081.mm.1 NR_045841.1 Refseq
          Mouse Expression Set 430 1453937_at - -
          Mouse Expression Set 430 1453937_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453937_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453937_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 650611 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0