Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02019477_s1
          See other CD209G GE Assays ›
          Gene Symbol:
          Cd209g
          Gene Name:
          CD209g antigen
          Gene Aliases:
          2310066I10Rik, AI481039, SIGNR7
          Chromosome Location:
          Chr.8: 4134246 - 4137707 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02019477_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd209g
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027343.3 NP_081619.3 8 750 176
          GenBank mRNA AK010071.1 - 7 711 176
          BC118542.1 - 1 3275 176

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P128636 BC118542.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P128636 AK010071.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P128636 AM286437.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P128636 NM_027343.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 52296 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P128636 BC118542.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P128636 AK010071.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P128636 AM286437.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P128636 NM_027343.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287436 AK010071.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137314 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128636 BC118542.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128636 AK010071.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128636 AM286437.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128636 NM_027343.2 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06832356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07192014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06863705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06890998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616274 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10569897 - -
          Affymetrix HT MG-430 PM Array Plate 1449918_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10569896 - -
          Mouse Genome 430 2.0 Array 1449918_PM_at - -
          Mouse Genome 430 2.0 Array 1449918_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70192_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70192_at - -
          Mouse Expression Set 430 1449918_PM_at - -
          Mouse Expression Set 430 1449918_at - -
          Mouse Genome 430A 2.0 Array 1449918_at - -
          Mouse Genome 430A 2.0 Array 1449918_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449918_at - -
          Affymetrix HT Mouse Genome 430A Array 1449918_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 312581 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1770403 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001770403 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1244759 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244759 - -
          mouse-6 v1.1 (Illumina) scl34122.7.1_24-S - -
          Illumina Sentrix mouse-6.v1 bead array 1770403 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl34122.7.1_24-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1770403 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2633260 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0