Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000014172.mm.1 |
TC1000001990.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000014172.mm.1 |
NR_015526.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014172.mm.1 |
NR_015526.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014175.mm.1 |
TC1000001990.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000014175.mm.1 |
NR_015526.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014175.mm.1 |
NR_015526.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014173.mm.1 |
TC1000001990.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000014173.mm.1 |
NR_015526.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014173.mm.1 |
NR_015526.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014174.mm.1 |
TC1000001990.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000014174.mm.1 |
NR_015526.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014174.mm.1 |
NR_015526.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014170.mm.1 |
TC1000001990.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000014170.mm.1 |
NR_015526.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014170.mm.1 |
NR_015526.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014171.mm.1 |
TC1000001990.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000014171.mm.1 |
NR_015526.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014171.mm.1 |
NR_015526.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014169.mm.1 |
TC1000001990.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000014169.mm.1 |
NR_015526.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014169.mm.1 |
NR_015526.2 |
Refseq |