Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02024514_s1
          See other 9530047P18RIK GE Assays ›
          Gene Symbol:
          9530047P18Rik
          Gene Name:
          RIKEN cDNA 9530047P18 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 65465797 - 65467093 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02024514_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9530047P18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK020597.1 - 2 1062 109

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream prepronociceptin Pnoc N/OFQ,
          N23K,
          Npnc1,
          OFQ/N
          -40.4kb Anti-sense Chr. 14 - 65400673 - 65425581 on NCBI Build GRCm38
          Downstream elongator acetyltransferase complex subunit 3 Elp3 2610507P14Rik,
          KAT9
          63.3kb Anti-sense Chr. 14 - 65530446 - 65593112 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2274996 AK020597.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11222864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11488435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11747221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11311343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11214891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11747219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11747215 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11811761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11246330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673704 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673661 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11315919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11341303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11817176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29673703 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432213_PM_at - -
          Mouse Genome 430 2.0 Array 1432213_PM_at - -
          Mouse Genome 430 2.0 Array 1432213_at - -
          Mouse Expression Set 430 1432213_PM_at - -
          Mouse Expression Set 430 1432213_at - -
          Affymetrix HT Mouse Genome 430B Array 1432213_at - -
          Affymetrix HT Mouse Genome 430B Array 1432213_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 440109 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103520358 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0