Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02025296_s1
          See other LOC433227 GE Assays ›
          Gene Symbol:
          LOC433227
          Gene Name:
          uncharacterized LOC433227
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02025296_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC433227
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK036283.1 - 2 3138 96

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream transducin-like enhancer of split 4,
          homolog of Drosophila E(spl)
          Tle4 5730411M05Rik,
          AA792082,
          Bce-1,
          Bce1,
          ESTM13,
          ESTM14,
          Grg4,
          X83332,
          X83333,
          grg-4
          -1.2mb Anti-sense Chr. 19 - 14448072 - 14598183 on NCBI Build GRCm38
          Downstream phosphoserine aminotransferase 1 Psat1 D8Ertd814e,
          EPIP,
          PSA,
          Psat
          97.5kb Anti-sense Chr. 19 - 15905123 - 15925059 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 254390 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 83022 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125854 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125854 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106639 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31976578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31976579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31976584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14364870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31976512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31976514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31976587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14375308 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31976516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14248300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31976593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31976596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31976599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31976598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14248299 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31976589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31976510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31976573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31976586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14512361 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31976576 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 825775 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0