Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02026230_g1
          See other SLC39A1 GE Assays ›
          Gene Symbol:
          Slc39a1
          Gene Name:
          solute carrier family 39 (zinc transporter), member 1
          Gene Aliases:
          Zip1, Zirtl
          Chromosome Location:
          Chr.3: 90248192 - 90253613 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02026230_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc39a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013901.2 NP_038929.2 4 1159 149
          GenBank mRNA AK146175.1 - 4 1151 149
          AK159290.1 - 4 1016 149
          BC005474.1 - 4 1121 3591856 149
          BC132461.1 - 3 1096 40130738 149
          BC138459.1 - 3 1096 8861479 149

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000785.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc39a1

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          transport
          ion transport
          zinc ion transport
          metal ion transport
          embryonic cranial skeleton morphogenesis
          transmembrane transport
          limb development
          zinc ion transmembrane transport
          signaling receptor binding
          zinc ion transmembrane transporter activity
          metal ion transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22324142 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098558 BC005474.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098558 NM_013901.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098558 AK159290.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 675164 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 723379 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 35565 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102632 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098558 BC005474.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098558 NM_013901.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098558 AK159290.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P106868 BC005474.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P106868 NM_013901.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P106868 AK159290.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P257023 Z78150.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324158 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324159 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102632 - -
          Whole Mouse Genome G4122A A_51_P106868 BC005474.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P106868 NM_013901.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P106868 AK159290.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P257023 Z78150.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119500 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121942 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129187 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129831 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102632 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02389586 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22324142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324251 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324158 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02989976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02989974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324145 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324137 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02892168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324152 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324165 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324134 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02389591 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324149 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324143 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02892170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324147 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02636847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02842412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324161 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324157 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10470949 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493710 - -
          Affymetrix HT MG-430 PM Array Plate 1424424_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10493709 - -
          Affymetrix Mouse Gene 1.1 ST Array 10470948 - -
          Mouse Genome 430 2.0 Array 1424424_at - -
          Mouse Genome 430 2.0 Array 1424424_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 30791_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 30791_at - -
          Affymetrix Mouse Clariom S Array PSR0300006103.mm.1 TC0300000785.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006103.mm.1 NM_013901.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006103.mm.1 NM_013901.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006106.mm.1 TC0300000785.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006106.mm.1 NM_013901.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006106.mm.1 NM_013901.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006100.mm.1 TC0300000785.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006100.mm.1 NM_013901.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006100.mm.1 NM_013901.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006102.mm.1 TC0300000785.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006102.mm.1 NM_013901.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006102.mm.1 NM_013901.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006101.mm.1 TC0300000785.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006101.mm.1 NM_013901.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006101.mm.1 NM_013901.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006104.mm.1 TC0300000785.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006104.mm.1 NM_013901.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006104.mm.1 NM_013901.2 Refseq
          Mouse Expression Set 430 1424424_PM_at - -
          Mouse Expression Set 430 1424424_at - -
          Murine Genome U74v2 Set 96832_PM_at - -
          Murine Genome U74v2 Set 100373_PM_at - -
          Murine Genome U74v2 Set 96832_at - -
          Murine Genome U74v2 Set 100373_at - -
          Mouse Genome 430A 2.0 Array 1424424_at - -
          Mouse Genome 430A 2.0 Array 1424424_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 553861 - -
          ABI Mouse Genome Survey Microarray 810533 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2570180 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002570180 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2616872 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2616872 - -
          mouse-6 v1.1 (Illumina) scl0030791.1_194-S - -
          Illumina Sentrix mouse-6.v1 bead array 2570180 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0030791.1_194-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2570180 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2616877 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2616877 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0030791.1_194-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0