Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02342625_g1
          See other RPL3 GE Assays ›
          Gene Symbol:
          Rpl3
          Gene Name:
          ribosomal protein L3
          Gene Aliases:
          F2, J1
          Chromosome Location:
          Chr.15: 80077781 - 80083406 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02342625_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013762.2 NP_038790.2 3 - 4 446 106
          GenBank mRNA AK088993.1 - 3 - 4 393 106
          AK144518.1 - 3 - 4 393 106
          AK144816.1 - 3 - 4 393 106
          AK146502.1 - 3 - 4 393 106
          AK149494.1 - 3 - 4 393 106
          AK151057.1 - 3 - 4 393 106
          AK151084.1 - 3 - 4 393 106
          AK151142.1 - 3 - 4 393 106
          AK151426.1 - 3 - 4 393 106
          AK151596.1 - 3 - 4 393 106
          AK151740.1 - 3 - 4 393 106
          AK166433.1 - 3 - 4 389 106
          AK166825.1 - 3 - 4 393 106
          AK167787.1 - 3 - 4 393 106
          AK167947.1 - 3 - 4 393 106
          AK168040.1 - 3 - 4 393 106
          AK168059.1 - 3 - 4 392 106
          AK168126.1 - 3 - 4 393 106
          AK168128.1 - 3 - 4 393 106
          AK168224.1 - 3 - 4 393 106
          AK168744.1 - 3 - 4 391 106
          AK169060.1 - 3 - 4 393 106
          AK169235.1 - 3 - 4 393 106
          AK169321.1 - 3 - 4 393 106
          AK170819.1 - 3 - 4 393 106
          AK172280.1 - 3 - 4 393 106
          AK192605.1 - 1 - 2 69 106
          BC009655.1 - 3 - 4 389 3485069 106
          BC083134.1 - 2 - 3 375 6504713 106
          BC085495.1 - 3 - 4 382 106
          BC094059.1 - 2 - 3 374 3676103 106
          BC158039.1 - 3 - 4 393 9007342 106
          Y00225.1 - 2 - 3 375 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001896.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpl3

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          cellular response to interleukin-4
          RNA binding
          structural constituent of ribosome

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1500014940.mm.1 TC1500001896.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500014940.mm.1 NM_013762.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014940.mm.1 NM_013762.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014941.mm.1 TC1500001896.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500014941.mm.1 NM_013762.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014941.mm.1 NM_013762.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014943.mm.1 TC1500001896.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500014943.mm.1 NM_013762.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014943.mm.1 NM_013762.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0