Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02342770_g1
          See other ACTR3 GE Assays ›
          Gene Symbol:
          Actr3
          Gene Name:
          ARP3 actin-related protein 3
          Gene Aliases:
          1200003A09Rik, Arp3
          Chromosome Location:
          Chr.1: 125392905 - 125435727 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02342770_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Actr3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001205385.1 NP_001192314.1 8 - 9 894 142
          NM_001205386.1 NP_001192315.1 8 - 9 891 142
          NM_023735.2 NP_076224.1 7 - 8 1086 142
          GenBank mRNA AK004554.1 - 7 - 8 900 142
          AK083343.1 - 7 - 8 904 142
          AK145536.1 - 7 - 8 906 142
          AK150487.1 - 7 - 8 788 142
          AK150589.1 - 7 - 8 864 142
          AK150991.1 - 7 - 8 844 142
          AK152865.1 - 7 - 8 881 142
          AK153079.1 - 7 - 8 907 142
          AK153150.1 - 7 - 8 881 142
          AK153204.1 - 7 - 8 787 142
          AK159135.1 - 7 - 8 873 142
          AK159668.1 - 7 - 8 798 142
          AK159775.1 - 7 - 8 885 142
          AK160047.1 - 7 - 8 746 142
          AK166529.1 - 8 - 9 838 142
          AK166586.1 - 8 - 9 838 142
          AK167148.1 - 7 - 8 845 142
          AK167891.1 - 7 - 8 843 142
          AK168568.1 - 7 - 8 773 142
          AK168771.1 - 7 - 8 859 142
          AK187649.1 - 2 - 3 207 142
          AK211935.1 - 3 - 4 324 142
          BC005557.1 - 7 - 8 849 3156748 142
          BC080806.1 - 8 - 9 852 5350752 142
          BC108386.1 - 7 - 8 817 5051125 142

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002993.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Actr3

          Gene Ontology Categories:

          Function(s) Process(es)

          actin filament organization
          establishment or maintenance of cell polarity
          asymmetric cell division
          positive regulation of lamellipodium assembly
          meiotic chromosome movement towards spindle pole
          cell projection organization
          positive regulation of actin filament polymerization
          meiotic cytokinesis
          Arp2/3 complex-mediated actin nucleation
          regulation of myosin II filament organization
          positive regulation of neuron differentiation
          positive regulation of dendrite morphogenesis
          meiotic cell cycle
          positive regulation of filopodium assembly
          spindle localization
          cilium assembly
          nucleotide binding
          actin binding
          structural constituent of cytoskeleton
          protein binding
          ATP binding
          actin filament binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100024041.mm.1 TC0100002993.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100024041.mm.1 NM_001205385.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100024041.mm.1 NM_001205385.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100024041.mm.1 NM_001205386.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100024041.mm.1 NM_001205386.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100024041.mm.1 NM_023735.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100024041.mm.1 NM_023735.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0