Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02391143_m1
          See other PISD-PS3 GE Assays ›
          Gene Symbol:
          Pisd-ps3
          Gene Name:
          phosphatidylserine decarboxylase, pseudogene 3
          Gene Aliases:
          4933439C20Rik
          Chromosome Location:
          Chr.Un|NT_187064: 52674 - 59689 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02391143_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pisd-ps3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003518.2 2 - 3 363 91
          GenBank mRNA AK005333.1 - 467 91
          AK017116.1 - 2 - 3 372 91
          AK034517.1 - 2 - 3 1020 91
          AK081907.1 - 2 - 3 340 91
          AK161130.1 - 2 - 3 363 91
          AK162734.1 - 2 - 3 1496 91
          AK168892.1 - 2 - 3 340 91
          AK169597.1 - 1 - 2 1106 91
          AK169755.1 - 1 - 2 1106 91
          BC059938.1 - 1 - 2 269 91
          BC083061.1 - 1 - 2 111 91
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CA319555.1 - 2 - 3 380 Chr. Un|NT_187064.1 - 55497 - 59619 [-] on Build GRCm38 91
          GenBankmRNA AU080934.1 - 378 Chr. - - [-] on Build 91
          GenBankmRNA BQ945049.1 - 1 - 2 268 Chr. Un|NT_187064.1 - 55117 - 58837 [-] on Build GRCm38 91
          GenBankmRNA CN672583.1 - 1 - 2 116 Chr. Un|NT_187064.1 - 55456 - 58688 [-] on Build GRCm38 91
          GenBankmRNA CX202339.1 - 2 - 3 Chr. Un|NT_187064.1 - 55209 - 58685 [+] on Build GRCm38 91
          GenBankmRNA CJ171901.1 - 2 - 3 336 Chr. Un|NT_187064.1 - 57106 - 59663 [-] on Build GRCm38 91
          GenBankmRNA BY739907.1 - 2 - 3 340 Chr. Un|NT_187064.1 - 55575 - 59667 [-] on Build GRCm38 91
          GenBankmRNA BB619177.1 - 2 - 3 340 Chr. Un|NT_187064.1 - 55615 - 59667 [-] on Build GRCm38 91
          GenBankmRNA BY255235.1 - 2 - 3 338 Chr. Un|NT_187064.1 - 57004 - 59667 [-] on Build GRCm38 91
          GenBankmRNA BQ745548.1 - 2 - 3 308 Chr. Un|NT_187064.1 - 55444 - 59637 [-] on Build GRCm38 91
          GenBankmRNA CA531192.1 - 1 - 2 83 Chr. Un|NT_187064.1 - 55449 - 58655 [-] on Build GRCm38 91
          GenBankmRNA CJ175330.1 - 2 - 3 341 Chr. Un|NT_187064.1 - 57039 - 59667 [-] on Build GRCm38 91
          GenBankmRNA CA318015.1 - 1 - 2 116 Chr. Un|NT_187064.1 - 54728 - 58687 [-] on Build GRCm38 91
          GenBankmRNA BB649681.1 - 2 - 3 340 Chr. Un|NT_187064.1 - 55570 - 59667 [-] on Build GRCm38 91
          GenBankmRNA BF140154.1 - 103 Chr. - - [-] on Build 92
          GenBankmRNA BQ443975.1 - 1 - 2 114 Chr. Un|NT_187064.1 - 54776 - 58677 [-] on Build GRCm38 91
          GenBankmRNA BP766142.1 - 1 - 2 451 Chr. Un|NT_187064.1 - 25388 - 58997 [-] on Build GRCm38 91
          GenBankmRNA CA533423.1 - 1 - 2 191 Chr. Un|NT_187064.1 - 57025 - 58768 [-] on Build GRCm38 91
          GenBankmRNA BY273730.1 - 2 - 3 341 Chr. Un|NT_187064.1 - 57051 - 59667 [-] on Build GRCm38 91
          GenBankmRNA AU066585.1 - 2 - 3 357 Chr. 11 - 3124028 - 3127613 [+] on Build GRCm38 91
          GenBankmRNA CX207012.1 - 2 - 3 362 Chr. Un|NT_187064.1 - 55530 - 59690 [-] on Build GRCm38 91
          GenBankmRNA CK620355.1 - 1 - 2 130 Chr. Un|NT_187064.1 - 53068 - 58702 [-] on Build GRCm38 91
          GenBankmRNA BI697592.1 - 1 - 2 114 Chr. Un|NT_187064.1 - 55243 - 58686 [-] on Build GRCm38 91
          GenBankmRNA CN671160.1 - 1 - 2 174 Chr. Un|NT_187064.1 - 55467 - 58748 [-] on Build GRCm38 91
          GenBankmRNA BM951557.1 - 1 - 2 110 Chr. Un|NT_187064.1 - 55277 - 58681 [-] on Build GRCm38 91
          GenBankmRNA CN681062.1 - 1 - 2 114 Chr. Un|NT_187064.1 - 25151 - 58686 [-] on Build GRCm38 91
          GenBankmRNA AA607577.1 - 1 - 2 87 Chr. Un|NT_187064.1 - 57059 - 58659 [-] on Build GRCm38 91
          GenBankmRNA CX237225.1 - 2 - 3 410 Chr. Un|NT_187064.1 - 55545 - 59731 [-] on Build GRCm38 91
          GenBankmRNA BY722385.1 - 2 - 3 335 Chr. Un|NT_187064.1 - 25366 - 59664 [-] on Build GRCm38 91
          GenBankmRNA BQ443910.1 - 1 - 2 255 Chr. Un|NT_187064.1 - 55206 - 58826 [-] on Build GRCm38 91
          GenBankmRNA BY716795.1 - 2 - 3 366 Chr. Un|NT_187064.1 - 56853 - 59695 [-] on Build GRCm38 91
          GenBankmRNA BU611278.1 - 2 - 3 477 Chr. Un|NT_187064.1 - 55658 - 59729 [-] on Build GRCm38 91
          GenBankmRNA CN680240.1 - 1 - 2 114 Chr. Un|NT_187064.1 - 55481 - 58686 [-] on Build GRCm38 91
          GenBankmRNA CB519629.1 - 292 Chr. - - [-] on Build 91
          GenBankmRNA CK618982.1 - 2 - 3 285 Chr. Un|NT_187064.1 - 57030 - 59613 [-] on Build GRCm38 91
          GenBankmRNA CF537165.1 - 119 Chr. - - [-] on Build 91
          GenBankmRNA CA315346.1 - 2 - 3 481 Chr. Un|NT_187064.1 - 55552 - 59792 [-] on Build GRCm38 91
          GenBankmRNA CJ174094.1 - 2 - 3 341 Chr. Un|NT_187064.1 - 57068 - 59667 [-] on Build GRCm38 91
          GenBankmRNA BP766143.1 - Chr. - - [-] on Build 91
          GenBankmRNA BF141965.1 - 102 Chr. - - [-] on Build 91
          GenBankmRNA CB245085.1 - 135 Chr. - - [-] on Build 91
          GenBankmRNA AW321782.1 - 1 - 2 113 Chr. Un|NT_187064.1 - 55453 - 58685 [-] on Build GRCm38 91

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream phosphatidylserine decarboxylase,
          pseudogene 1
          Pisd-ps1 Sense Chr. 11 - 3124021 - 3131945 on NCBI Build GRCm38
          Downstream Sfi1 homolog,
          spindle assembly associated (yeast)
          Sfi1 4.2kb Anti-sense Chr. 11 - 3131850 - 3203343 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TCUn_JH58430400000005.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSRUn_JH58430400000022.mm.1 TCUn_JH58430400000005.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSRUn_JH58430400000022.mm.1 NR_003518.2 Refseq
          Affymetrix Mouse Clariom S Array PSRUn_JH58430400000025.mm.1 TCUn_JH58430400000005.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSRUn_JH58430400000025.mm.1 NR_003518.2 Refseq
          Affymetrix Mouse Clariom S Array PSRUn_JH58430400000023.mm.1 TCUn_JH58430400000005.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSRUn_JH58430400000023.mm.1 NR_003518.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 653407 - -
          ABI Mouse Genome Survey Microarray 711766 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0