Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02524711_g1
          See other RPL41 GE Assays ›
          Gene Symbol:
          Rpl41
          Gene Name:
          ribosomal protein L41
          Gene Aliases:
          1810055P16Rik, 2210411K19Rik
          Chromosome Location:
          Chr.10: 128548110 - 128549168 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02524711_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl41
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_018860.4 NP_061348.1 4 180 72
          GenBank mRNA AK002503.1 - 4 126 72
          AK007874.1 - 4 128 72
          AK009548.1 - 4 127 72
          AK010246.1 - 4 127 72
          AK010317.1 - 4 127 72
          AK010482.1 - 4 127 72
          AK010509.1 - 4 127 72
          AK010564.1 - 4 125 72
          AK010583.1 - 4 127 72
          AK010585.1 - 4 125 72
          AK012252.1 - 4 125 72
          AK012450.1 - 4 130 72
          AK012476.1 - 4 126 72
          AK012487.1 - 4 125 72
          AK013324.1 - 4 132 72
          AK019049.1 - 4 127 72
          AK019178.1 - 4 122 72
          AK028126.1 - 4 125 72
          AK028231.1 - 4 128 72
          AK028263.1 - 4 125 72
          AK150502.1 - 4 127 72
          AK151566.1 - 4 126 72
          AK160588.1 - 4 127 72
          AK168107.1 - 4 127 72
          AK168485.1 - 4 125 72
          AK168653.1 - 4 125 72
          AK168719.1 - 4 125 72
          AV478133.1 - 4 113 72
          BC081450.1 - 3 196 72
          BC083066.1 - 4 114 72
          BC089572.1 - 4 124 72
          BC089573.1 - 4 126 72
          BC092010.1 - 4 115 72
          BC099380.1 - 3 194 72
          BC100592.1 - 4 126 72
          BC145727.1 - 4 127 40131030 72
          BC145729.1 - 4 127 40131032 72
          BU937034.1 - 4 183 72
          DQ113647.1 - 86 72
          U93862.1 - 3 101 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000003151.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpl41

          Gene Ontology Categories:

          Process(es)

          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27541975 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27541978 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27541977 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27541976 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027819 AK168485.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027819 AK168653.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027819 AK168719.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027819 AV478133.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027819 BC081450.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027819 AK168485.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027819 AK168653.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027819 AK168719.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027819 AV478133.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027819 BC081450.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P295201 U93862.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P295201 AK168485.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P295201 AK168653.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P295201 AK168719.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P295201 AV478133.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541987 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541979 - -
          Whole Mouse Genome G4122A A_52_P295201 U93862.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P295201 AK168485.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P295201 AK168653.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P295201 AK168719.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P295201 AV478133.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107150 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08592371 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27541977 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27541975 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27541978 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27541976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08592371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08807340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08592396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541971 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10373449 - -
          Affymetrix Mouse Gene 1.0 ST Array 10416916 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412099 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373450 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368929 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591868 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373451 - -
          Affymetrix HT MG-430 PM Array Plate 1422623_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1454639_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1455578_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1433530_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10412098 - -
          Affymetrix Mouse Gene 1.1 ST Array 10373448 - -
          Affymetrix Mouse Gene 1.1 ST Array 10416915 - -
          Affymetrix Mouse Gene 1.1 ST Array 10368928 - -
          Affymetrix Mouse Gene 1.1 ST Array 10591867 - -
          Mouse Genome 430 2.0 Array 1422623_x_at - -
          Mouse Genome 430 2.0 Array 1455578_x_at - -
          Mouse Genome 430 2.0 Array 1446726_at - -
          Mouse Genome 430 2.0 Array 1446726_PM_at - -
          Mouse Genome 430 2.0 Array 1422623_PM_x_at - -
          Mouse Genome 430 2.0 Array 1455578_PM_x_at - -
          Mouse Genome 430 2.0 Array 1454639_x_at - -
          Mouse Genome 430 2.0 Array 1454639_PM_x_at - -
          Affymetrix Mouse Clariom S Array PSR1000023673.mm.1 TC1000003151.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000023673.mm.1 NM_018860.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023673.mm.1 NM_018860.4 Refseq
          Mouse Expression Set 430 1454639_x_at - -
          Mouse Expression Set 430 1455578_PM_x_at - -
          Mouse Expression Set 430 1455578_x_at - -
          Mouse Expression Set 430 1454639_PM_x_at - -
          Mouse Expression Set 430 1422623_PM_x_at - -
          Mouse Expression Set 430 1422623_x_at - -
          Mouse Expression Set 430 1446726_PM_at - -
          Mouse Expression Set 430 1446726_at - -
          Murine Genome U74v2 Set 100213_PM_f_at - -
          Murine Genome U74v2 Set 100213_f_at - -
          Murine Genome U74v2 Set 169735_PM_at - -
          Murine Genome U74v2 Set 169735_at - -
          Murine Genome U74v2 Set 140477_PM_f_at - -
          Murine Genome U74v2 Set 140477_f_at - -
          Mouse Genome 430A 2.0 Array 1454639_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1422623_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1454639_x_at - -
          Mouse Genome 430A 2.0 Array 1455578_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1455578_x_at - -
          Mouse Genome 430A 2.0 Array 1422623_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1455578_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1422623_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1455578_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1422623_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1446726_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1446726_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 900483 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6940112 - -
          Illumina mouse-6 v1.0 expression beadchip 6940112 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006940112 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2620695 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2620695 - -
          mouse-6 v1.1 (Illumina) scl067945.1_102-S - -
          Illumina Sentrix mouse-6.v1 bead array 6940112 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067945.1_102-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6940112 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2620695 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2620695 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl067945.1_102-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline