Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02525306_s1
          See other TMEM185B GE Assays ›
          Gene Symbol:
          Tmem185b
          Gene Name:
          transmembrane protein 185B
          Gene Aliases:
          2500001K11Rik
          Chromosome Location:
          Chr.1: 119526154 - 119528983 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02525306_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem185b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146103.2 NP_666215.1 1 1961 118
          GenBank mRNA AK010846.1 - 1 1677 118
          AK031130.1 - 1 1956 118
          BC020408.1 - 1 1949 118
          BC024712.1 - 1 1948 2646072 118
          BC051523.1 - 1 818 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001076.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem185b

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20557522 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20557525 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20557524 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20557527 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20557526 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2184925 BC024712.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2184925 AK010846.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2184925 AK031130.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2184925 BC020408.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2184925 NM_146103.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 796482 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154874 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2184925 BC024712.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2184925 AK010846.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2184925 AK031130.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2184925 BC020408.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2184925 NM_146103.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20557499 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154874 - -
          Whole Mouse Genome G4122A A_52_P411655 BC024712.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P411655 BC051523.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P411655 AK031130.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P411655 BC020408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P411655 NM_146103.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115814 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116365 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154874 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00631581 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00848787 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00848784 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00176865 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00848775 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00631580 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00848761 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20557525 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20557527 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20557524 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20557522 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20557526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20557524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00285843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01108514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00848787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00622855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00906002 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00285845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557494 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557544 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00631580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00848784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557517 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00176865 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00259743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557571 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00896703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557556 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00896711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00324841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557572 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20557505 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10349317 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349318 - -
          Affymetrix HT MG-430 PM Array Plate 1452093_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10349316 - -
          Mouse Genome 430 2.0 Array 1452093_at - -
          Mouse Genome 430 2.0 Array 1452093_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 226351_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 226351_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100008843.mm.1 TC0100001076.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100008843.mm.1 NM_146103.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100008843.mm.1 NM_146103.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100008846.mm.1 TC0100001076.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100008846.mm.1 NM_146103.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100008846.mm.1 NM_146103.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100008845.mm.1 TC0100001076.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100008845.mm.1 NM_146103.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100008845.mm.1 NM_146103.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100008849.mm.1 TC0100001076.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100008849.mm.1 NM_146103.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100008849.mm.1 NM_146103.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100008844.mm.1 TC0100001076.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100008844.mm.1 NM_146103.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100008844.mm.1 NM_146103.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100008847.mm.1 TC0100001076.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100008847.mm.1 NM_146103.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100008847.mm.1 NM_146103.2 Refseq
          Mouse Expression Set 430 1452093_at - -
          Mouse Expression Set 430 1452093_PM_at - -
          Murine Genome U74v2 Set 168485_at - -
          Murine Genome U74v2 Set 166076_r_at - -
          Murine Genome U74v2 Set 166076_PM_r_at - -
          Murine Genome U74v2 Set 168485_PM_at - -
          Mouse Genome 430A 2.0 Array 1452093_at - -
          Mouse Genome 430A 2.0 Array 1452093_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452093_at - -
          Affymetrix HT Mouse Genome 430A Array 1452093_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 313142 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5080577 - -
          MouseWG-6 v2 Expression BeadChip 104920647 - -
          Illumina mouse-6 v1.0 expression beadchip 104920647 - -
          Illumina mouse-6 v1.0 expression beadchip 5080577 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005080577 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2696272 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2696272 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2483887 - -
          mouse-6 v1.1 (Illumina) scl17559.2_427-S - -
          mouse-6 v1.1 (Illumina) scl00226351.2_117-S - -
          Illumina Sentrix mouse-6.v1 bead array 5080577 - -
          Illumina Sentrix mouse-6.v1 bead array 104920647 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00226351.2_117-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5080577 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104920647 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2696272 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2696272 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl00226351.2_117-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6bbccd986b-mx52t:80/100.66.130.62:80.
          git-commit: 717389b07c2b9f75290ff02e811d41bb86d4254a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0