Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02581461_m1
          See other TMEM181A GE Assays ›
          Gene Symbol:
          Tmem181a Tmem181b-ps Tmem181c-ps
          Gene Name:
          transmembrane protein 181A
          transmembrane protein 181B, pseudogene
          transmembrane protein 181C, pseudogene
          Gene Aliases:
          5930418K15Rik, C76977, Gpr178, Tmem181, mKIAA1423
          Tmem181b
          100040596, ENSMUSG00000079729, ENSMUSG00000079731, Tmem181d-ps
          Chromosome Location:
          Chr.17: 6256865 - 6308314 on Build GRCm38
          Chromosome Location:
          Chr.17: 6439002 - 6450994 on Build GRCm38
          Chromosome Location:
          Chr.17: 6610103 - 6620925 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Mm02581461_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem181a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033178.3 NP_001028350.1 15 - 16 1389 66
          GenBank mRNA AK122507.1 - 12 - 13 2657 66
          AK154554.1 - 15 - 16 1385 66
          AK208144.1 - 3 - 4 227 66
          AW825806.1 - 2 - 3 178 66
          Gene Symbol: Tmem181b-ps
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033520.1 8 - 9 2163 66
          GenBank mRNA AK039520.1 - 7 - 8 789 66
          AK137412.1 - 8 - 9 2163 66
          BC147312.1 - 1369 9056375 66
          BC147313.1 - 1369 9056366 66
          Gene Symbol: Tmem181c-ps
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028305.2 7 - 8 1051 66
          GenBank mRNA AK045744.1 - 7 - 8 1052 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000055.mm.1, TC1700001384.mm.1, TC1700001386.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem181a
          Click here to find siRNAs for Gene Tmem181b-ps
          Click here to find siRNAs for Gene Tmem181c-ps

          Gene Ontology Categories:

          Process(es)

          toxic substance binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700000379.mm.1 TC1700000055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000379.mm.1 NM_001033178.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000382.mm.1 TC1700000055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000382.mm.1 NM_001033178.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000385.mm.1 TC1700000055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000385.mm.1 NM_001033178.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012457.mm.1 TC1700001384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012457.mm.1 NR_033520.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012457.mm.1 NR_033520.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012462.mm.1 TC1700001384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012462.mm.1 NR_033520.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012462.mm.1 NR_033520.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012463.mm.1 TC1700001384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012463.mm.1 NR_033520.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012463.mm.1 NR_033520.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012464.mm.1 TC1700001384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012464.mm.1 NR_033520.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012464.mm.1 NR_033520.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012504.mm.1 TC1700001386.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012504.mm.1 NR_028305.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012468.mm.1 TC1700001384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012469.mm.1 TC1700001384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012471.mm.1 TC1700001384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000383.mm.1 TC1700000055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000383.mm.1 NM_001033178.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012452.mm.1 TC1700001384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012452.mm.1 NR_033520.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012452.mm.1 NR_033520.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012454.mm.1 TC1700001384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012454.mm.1 NR_033520.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012454.mm.1 NR_033520.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012459.mm.1 TC1700001384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012459.mm.1 NR_033520.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012459.mm.1 NR_033520.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000381.mm.1 TC1700000055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000381.mm.1 NM_001033178.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000388.mm.1 TC1700000055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000388.mm.1 NM_001033178.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012453.mm.1 TC1700001384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012453.mm.1 NR_033520.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012453.mm.1 NR_033520.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000378.mm.1 TC1700000055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000378.mm.1 NM_001033178.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000386.mm.1 TC1700000055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000386.mm.1 NM_001033178.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000387.mm.1 TC1700000055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000387.mm.1 NM_001033178.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000390.mm.1 TC1700000055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000390.mm.1 NM_001033178.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000391.mm.1 TC1700000055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000391.mm.1 NM_001033178.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012450.mm.1 TC1700001384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012450.mm.1 NR_033520.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012450.mm.1 NR_033520.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012503.mm.1 TC1700001386.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012503.mm.1 NR_028305.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000384.mm.1 TC1700000055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000384.mm.1 NM_001033178.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000389.mm.1 TC1700000055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000389.mm.1 NM_001033178.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000392.mm.1 TC1700000055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000392.mm.1 NM_001033178.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000393.mm.1 TC1700000055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000393.mm.1 NM_001033178.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012467.mm.1 TC1700001384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012473.mm.1 TC1700001384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012466.mm.1 TC1700001384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012466.mm.1 NR_033520.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012466.mm.1 NR_033520.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012472.mm.1 TC1700001384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000380.mm.1 TC1700000055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000380.mm.1 NM_001033178.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012449.mm.1 TC1700001384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012449.mm.1 NR_033520.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012449.mm.1 NR_033520.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012451.mm.1 TC1700001384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012451.mm.1 NR_033520.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012451.mm.1 NR_033520.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012460.mm.1 TC1700001384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012460.mm.1 NR_033520.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012460.mm.1 NR_033520.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012505.mm.1 TC1700001386.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012505.mm.1 NR_028305.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012470.mm.1 TC1700001384.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0