Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02581462_g1
          See other TMEM181A GE Assays ›
          Gene Symbol:
          Tmem181a Tmem181b-ps Tmem181c-ps
          Gene Name:
          transmembrane protein 181A
          transmembrane protein 181B, pseudogene
          transmembrane protein 181C, pseudogene
          Gene Aliases:
          5930418K15Rik, C76977, Gpr178, Tmem181, mKIAA1423
          Tmem181b
          100040596, ENSMUSG00000079729, ENSMUSG00000079731, Tmem181d-ps
          Chromosome Location:
          Chr.17: 6256865 - 6308314 on Build GRCm38
          Chromosome Location:
          Chr.17: 6439002 - 6450994 on Build GRCm38
          Chromosome Location:
          Chr.17: 6610103 - 6620925 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Mm02581462_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem181a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033178.3 NP_001028350.1 16 - 17 1460 102
          GenBank mRNA AK154554.1 - 16 - 17 1456 102
          AW825806.1 - 3 - 4 249 102
          Gene Symbol: Tmem181b-ps
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033520.1 9 - 10 2234 102
          GenBank mRNA AK039520.1 - 8 - 9 860 102
          AK137412.1 - 9 - 10 2234 102
          BC147312.1 - 1440 9056375 102
          BC147313.1 - 1440 9056366 102
          Gene Symbol: Tmem181c-ps
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028305.2 8 - 9 1122 102
          GenBank mRNA AK045744.1 - 8 - 9 1123 102
          BB309117.2 - 2 - 3 262 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000055.mm.1, TC1700001384.mm.1, TC1700001386.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem181a
          Click here to find siRNAs for Gene Tmem181b-ps
          Click here to find siRNAs for Gene Tmem181c-ps

          Gene Ontology Categories:

          Process(es)

          toxic substance binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021099 AK083137.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021099 AK137412.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021099 NR_033520.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021099 AK039520.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021099 AK154554.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021094 BC096660.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021094 BC032285.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 921355 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 66657 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 21535 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 15086 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021099 AK083137.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021099 AK137412.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021099 NR_033520.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021099 AK039520.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021099 AK154554.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021094 BC096660.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021094 BC032285.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29006763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29006803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29006797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29006806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29006796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29006804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29006788 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29006765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29006793 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29006792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29006799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29006790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29006770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29006795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29006789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29006768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29006769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29006801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29006767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29006786 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07861927 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09574250 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09579641 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09579647 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09579652 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09579628 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09579637 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07861937 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09579659 - -
          Whole Mouse Genome G4122A A_52_P503927 BC147312.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P503927 BC147313.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P503927 AK137412.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P503927 BC032285.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P503927 NR_033520.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09579657 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09579630 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09574250 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07861937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29006755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31022119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10914760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33002998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33003017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33003028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006747 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014243 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33002997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006756 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33003031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33003003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33003019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006701 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33003018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006720 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006742 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33003009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33003027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10636607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006718 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014245 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33003022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014231 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014222 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33003005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33003030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31022119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33003033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33003025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33003021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006693 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33003034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33003024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006745 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006765 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33003035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006750 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33003023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10551541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006738 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33014233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29006757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P33003026 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.1 ST Array 10441511 - -
          Affymetrix Mouse Gene 1.1 ST Array 10441511 - -
          Affymetrix Mouse Gene 1.1 ST Array 10441511 - -
          Mouse Genome 430 2.0 Array 1440862_PM_at - -
          Mouse Genome 430 2.0 Array 1435948_PM_at - -
          Mouse Genome 430 2.0 Array 1455748_PM_at - -
          Mouse Genome 430 2.0 Array 1435948_at - -
          Mouse Genome 430 2.0 Array 1455748_at - -
          Mouse Genome 430 2.0 Array 1455744_at - -
          Mouse Genome 430 2.0 Array 1428117_x_at - -
          Mouse Genome 430 2.0 Array 1440862_at - -
          Mouse Genome 430 2.0 Array 1452848_at - -
          Mouse Genome 430 2.0 Array 1455744_PM_at - -
          Mouse Genome 430 2.0 Array 1428117_PM_x_at - -
          Mouse Genome 430 2.0 Array 1452848_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700012499.mm.1 TC1700001386.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012499.mm.1 NR_028305.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012500.mm.1 TC1700001386.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012500.mm.1 NR_028305.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000394.mm.1 TC1700000055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000394.mm.1 NM_001033178.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000397.mm.1 TC1700000055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000397.mm.1 NM_001033178.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012448.mm.1 TC1700001384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012448.mm.1 NR_033520.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012448.mm.1 NR_033520.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012498.mm.1 TC1700001386.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012498.mm.1 NR_028305.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000395.mm.1 TC1700000055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000395.mm.1 NM_001033178.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012502.mm.1 TC1700001386.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012502.mm.1 NR_028305.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012501.mm.1 TC1700001386.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012501.mm.1 NR_028305.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012447.mm.1 TC1700001384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012447.mm.1 NR_033520.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012447.mm.1 NR_033520.1 Refseq
          Mouse Expression Set 430 1455748_PM_at - -
          Mouse Expression Set 430 1455748_at - -
          Mouse Expression Set 430 1435948_PM_at - -
          Mouse Expression Set 430 1452848_PM_at - -
          Mouse Expression Set 430 1435948_at - -
          Mouse Expression Set 430 1452848_at - -
          Mouse Expression Set 430 1435948_PM_at - -
          Mouse Expression Set 430 1435948_at - -
          Murine Genome U74v2 Set 102870_PM_at - -
          Murine Genome U74v2 Set 95958_at - -
          Murine Genome U74v2 Set 95958_PM_at - -
          Murine Genome U74v2 Set 102870_at - -
          Mouse Genome 430A 2.0 Array 1455748_PM_at - -
          Mouse Genome 430A 2.0 Array 1455748_at - -
          Affymetrix HT Mouse Genome 430B Array 1452848_at - -
          Affymetrix HT Mouse Genome 430B Array 1452848_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2744121 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245263 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2744121 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0