Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02581613_m1
          See other 2010002M12RIK GE Assays ›
          Gene Symbol:
          2010002M12Rik
          Gene Name:
          RIKEN cDNA 2010002M12 gene
          Gene Aliases:
          AW412491, Ifit1b
          Chromosome Location:
          Chr.19: 34617051 - 34640743 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02581613_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2010002M12Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053217.2 NP_444447.1 2 - 3 236 71
          GenBank mRNA AK008052.1 - 1 - 2 88 71
          AK136525.1 - 1 - 2 88 71
          AK153247.1 - 2 - 3 237 71
          BC138202.1 - 1 - 2 67 8861221 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001423.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK008052.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 2010002M12Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          response to virus
          intracellular transport of viral protein in host cell
          negative regulation of protein binding
          positive regulation of viral genome replication
          negative regulation of viral genome replication
          negative regulation of defense response to virus by host
          negative regulation of helicase activity
          cellular response to type I interferon
          cellular response to exogenous dsRNA
          RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030568 AK136525.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030568 AK153247.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030568 NM_053217.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 692731 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 44360 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 104780 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 648722 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 286636 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735667 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 770614 AK006596.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 770614 AK077003.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 770614 AK077188.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 770614 BC117739.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 770614 NM_028494.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 770614 XM_001472420.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 865231 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 442013 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 820607 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 722106 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 48063 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 149899 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146206 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126155 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166034 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030568 AK136525.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030568 AK153247.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030568 NM_053217.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32081345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32081351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32081349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32081343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32081352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32081340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32081342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32081348 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126155 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146206 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09158586 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166034 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09158565 - -
          Whole Mouse Genome G4122A A_52_P289972 BC003493.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P289972 BC138202.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P289972 AK136525.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P289972 AK153247.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P289972 NM_053217.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124356 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126155 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166034 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146206 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14249580 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14249584 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14249588 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14249583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081342 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14352713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081280 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14292255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14352710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081291 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14249583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14416504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14249584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14424475 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14235043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081327 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081292 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14292250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14416505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14249588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081276 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14292264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081342 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081332 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081352 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10467151 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467150 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467152 - -
          Affymetrix HT MG-430 PM Array Plate 1438311_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1457976_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10467149 - -
          Mouse Genome 430 2.0 Array 1438311_at - -
          Mouse Genome 430 2.0 Array 1457976_PM_at - -
          Mouse Genome 430 2.0 Array 1457976_at - -
          Mouse Genome 430 2.0 Array 1438311_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 112419_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 112419_at - -
          Affymetrix Mouse Clariom S Array PSR1900012442.mm.1 TC1900001423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900012442.mm.1 NM_053217.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012442.mm.1 NM_053217.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012441.mm.1 TC1900001423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900012441.mm.1 NM_053217.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012441.mm.1 NM_053217.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012440.mm.1 TC1900001423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900012440.mm.1 NM_053217.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012440.mm.1 NM_053217.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012445.mm.1 TC1900001423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900012445.mm.1 NM_053217.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012445.mm.1 NM_053217.2 Refseq
          Mouse Expression Set 430 1457976_PM_at - -
          Mouse Expression Set 430 1438311_PM_at - -
          Mouse Expression Set 430 1457976_at - -
          Mouse Expression Set 430 1438311_at - -
          Murine Genome U74v2 Set 135535_PM_i_at - -
          Murine Genome U74v2 Set 135536_r_at - -
          Murine Genome U74v2 Set 135536_PM_r_at - -
          Murine Genome U74v2 Set 135535_i_at - -
          Affymetrix HT Mouse Genome 430B Array 1457976_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457976_at - -
          Affymetrix HT Mouse Genome 430B Array 1438311_at - -
          Affymetrix HT Mouse Genome 430B Array 1438311_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 779621 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5420253 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2693141 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2693141 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0