Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02601036_g1
          See other DNAAF3 GE Assays ›
          Gene Symbol:
          Dnaaf3
          Gene Name:
          dynein, axonemal assembly factor 3
          Gene Aliases:
          6030429G01Rik, b2b1739Clo
          Chromosome Location:
          Chr.7: 4522948 - 4532488 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02601036_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnaaf3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033548.2 NP_001028720.1 11 - 12 1285 74
          GenBank mRNA AK134335.1 - 11 - 12 1286 74
          BC150983.1 - 11 - 12 1271 9087896 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004637.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          cell projection organization
          motile cilium assembly
          axonemal dynein complex assembly
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072284 AK134335.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072284 NM_001033548.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 890216 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 454480 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 352152 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 498516 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 199814 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 50121 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2072284 AK134335.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2072284 NM_001033548.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949288 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949279 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949271 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01475589 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06455444 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06455446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06257183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06584007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06173729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06339442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06455443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06452260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06453582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06285094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06592032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06592020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06592035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06161007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06335363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06357349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06592023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06589595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06592027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06636514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06455452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06285096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06592033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06548838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06285095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06592034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06455444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06592028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06590486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949327 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10559576 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559573 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559579 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559572 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559570 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559578 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559575 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559569 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559571 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559574 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559577 - -
          Affymetrix Mouse Gene 1.1 ST Array 10559568 - -
          Affymetrix Mouse Clariom S Array PSR0700019702.mm.1 TC0700004637.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700019702.mm.1 NM_001033548.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019702.mm.1 NM_001033548.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019701.mm.1 TC0700004637.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700019701.mm.1 NM_001033548.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019701.mm.1 NM_001033548.2 Refseq
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2959253 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2959253 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0