Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02601778_g1
          See other RPS18 GE Assays ›
          Gene Symbol:
          Rps18
          Gene Name:
          ribosomal protein S18
          Gene Aliases:
          H-2Ke3, H2-Ke3, Ke-3, ke3
          Chromosome Location:
          Chr.17: 33951999 - 33955641 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02601778_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rps18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011296.2 NP_035426.1 4 - 5 406 84
          GenBank mRNA AK050626.1 - 4 - 5 429 84
          AK144398.1 - 84
          AK146354.1 - 4 - 5 429 84
          AK151580.1 - 4 - 5 429 84
          AK159437.1 - 4 - 5 429 84
          AK159821.1 - 4 - 5 429 84
          AK169246.1 - 4 - 5 429 84
          AK169732.1 - 3 - 4 83
          BC081458.1 - 3 - 4 394 5712669 84
          BC081459.1 - 4 - 5 406 6407328 84
          BC091729.1 - 4 - 5 404 84
          BC093500.1 - 4 - 5 407 4207659 84
          BC100604.1 - 4 - 5 428 30120583 84
          M76763.1 - 4 - 5 407 84

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rps18

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          nucleic acid binding
          RNA binding
          structural constituent of ribosome
          protein binding
          rRNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31151238 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31151243 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31151240 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31151242 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31151239 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135631 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135632 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 875423 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 344841 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134800 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135632 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135631 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P193899 BC093500.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P193899 BC100604.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P193899 M76763.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P193899 BC091729.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P193899 AK146354.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P287677 BC093500.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P287677 BC100604.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P287677 M76763.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P287677 BC091729.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P287677 AK146354.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151228 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134800 - -
          Whole Mouse Genome G4122A A_51_P287677 BC093500.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287677 BC100604.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287677 M76763.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287677 BC091729.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287677 AK146354.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P274544 - -
          Whole Mouse Genome G4122A A_52_P193899 BC093500.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P193899 BC100604.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P193899 M76763.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P193899 BC091729.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P193899 AK146354.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134800 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13178482 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13178486 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13178495 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13178489 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13434055 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13178484 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13434083 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31151242 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31151238 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31151240 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31151243 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31151239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253403 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31151242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31151219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31151232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10847966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253442 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253380 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11004205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253418 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10847967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253395 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13178493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11011945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253423 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13178476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31151229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11102595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31151221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13178486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253437 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253406 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31151236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31151225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253438 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253416 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253377 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31151240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10649899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253413 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253444 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253439 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253425 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10563464 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253434 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31151228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13178482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10595208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13338699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11099772 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31151227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29253398 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450067 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549803 - -
          Affymetrix Mouse Gene 1.0 ST Array 10513879 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450064 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409969 - -
          Affymetrix Mouse Gene 1.0 ST Array 10411394 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450065 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581536 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404355 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450068 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581537 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450066 - -
          Affymetrix HT MG-430 PM Array Plate 1421837_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1455572_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1421838_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1450390_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10450063 - -
          Affymetrix Mouse Gene 1.1 ST Array 10549802 - -
          Affymetrix Mouse Gene 1.1 ST Array 10404354 - -
          Affymetrix Mouse Gene 1.1 ST Array 10513878 - -
          Affymetrix Mouse Gene 1.1 ST Array 10411393 - -
          Affymetrix Mouse Gene 1.1 ST Array 10581535 - -
          Affymetrix Mouse Gene 1.1 ST Array 10409968 - -
          Mouse Genome 430 2.0 Array 1435712_a_at - -
          Mouse Genome 430 2.0 Array 1421838_PM_at - -
          Mouse Genome 430 2.0 Array 1421837_at - -
          Mouse Genome 430 2.0 Array 1450390_x_at - -
          Mouse Genome 430 2.0 Array 1448739_x_at - -
          Mouse Genome 430 2.0 Array 1421837_PM_at - -
          Mouse Genome 430 2.0 Array 1448739_PM_x_at - -
          Mouse Genome 430 2.0 Array 1421838_at - -
          Mouse Genome 430 2.0 Array 1450390_PM_x_at - -
          Mouse Genome 430 2.0 Array 1455572_PM_x_at - -
          Mouse Genome 430 2.0 Array 1455572_x_at - -
          Mouse Genome 430 2.0 Array 1435712_PM_a_at - -
          Affymetrix Mouse Clariom S Array PSR1700017534.mm.1 NM_011296.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017534.mm.1 NM_011296.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017534.mm.1 TC1700001888.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700017536.mm.1 NM_011296.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017536.mm.1 NM_011296.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017536.mm.1 TC1700001888.mm.1 MTA TC
          Mouse Expression Set 430 1435712_PM_a_at - -
          Mouse Expression Set 430 1421838_at - -
          Mouse Expression Set 430 1421837_PM_at - -
          Mouse Expression Set 430 1448739_PM_x_at - -
          Mouse Expression Set 430 1421838_PM_at - -
          Mouse Expression Set 430 1448739_x_at - -
          Mouse Expression Set 430 1421837_at - -
          Mouse Expression Set 430 1455572_PM_x_at - -
          Mouse Expression Set 430 1435712_a_at - -
          Mouse Expression Set 430 1455572_x_at - -
          Mouse Expression Set 430 1450390_x_at - -
          Mouse Expression Set 430 1450390_PM_x_at - -
          Murine Genome U74v2 Set 98333_at - -
          Murine Genome U74v2 Set 98333_PM_at - -
          Mouse Genome 430A 2.0 Array 1435712_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1455572_x_at - -
          Mouse Genome 430A 2.0 Array 1455572_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1421838_PM_at - -
          Mouse Genome 430A 2.0 Array 1448739_x_at - -
          Mouse Genome 430A 2.0 Array 1435712_a_at - -
          Mouse Genome 430A 2.0 Array 1450390_x_at - -
          Mouse Genome 430A 2.0 Array 1448739_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1421837_PM_at - -
          Mouse Genome 430A 2.0 Array 1450390_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1421837_at - -
          Mouse Genome 430A 2.0 Array 1421838_at - -
          Affymetrix HT Mouse Genome 430A Array 1421838_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421837_at - -
          Affymetrix HT Mouse Genome 430A Array 1421837_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450390_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1421838_at - -
          Affymetrix HT Mouse Genome 430A Array 1450390_x_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2030722 - -
          MouseWG-6 v2 Expression BeadChip 670132 - -
          Illumina mouse-6 v1.0 expression beadchip 2030722 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002030722 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2639750 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2639750 - -
          mouse-6 v1.1 (Illumina) scl0001735.1_49-S - -
          Illumina Sentrix mouse-6.v1 bead array 2030722 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001735.1_49-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2030722 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2639750 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0001735.1_49-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0