Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02602707_g1
          See other TIMM8A1 GE Assays ›
          Gene Symbol:
          Timm8a1
          Gene Name:
          translocase of inner mitochondrial membrane 8A1
          Gene Aliases:
          DXHXS1274E, Ddp1, Fci-12, Tim8a, Timm8a
          Chromosome Location:
          Chr.X: 134537258 - 134541629 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02602707_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Timm8a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013898.2 NP_038926.1 1 - 2 171 108
          GenBank mRNA AB031055.1 - 1 - 2 171 108
          AF150081.1 - 1 - 2 162 108
          AK011402.1 - 1 - 2 206 108
          AK088903.1 - 1 - 2 164 108
          AK090079.1 - 1 - 2 210 108
          AK165573.1 - 1 - 2 166 108
          BC004796.1 - 1 - 2 159 3588797 108
          BC094631.1 - 1 - 2 180 3419065 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00002993.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Timm8a1

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting to mitochondrion
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672450 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672455 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672454 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672453 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672451 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964302 BC004796.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952744 BC094631.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952744 AK165573.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952744 BC004796.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952744 AB031055.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952744 NM_013898.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858538 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 138592 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167291 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952744 BC094631.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952744 AK165573.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952744 BC004796.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952744 AB031055.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952744 NM_013898.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964302 BC004796.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P373043 BC094631.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P373043 AK165573.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P373043 BC004796.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P373043 AB031055.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P373043 NM_013898.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672457 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167291 - -
          Whole Mouse Genome G4122A A_51_P373043 BC094631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P373043 AK165573.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P373043 BC004796.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P373043 AB031055.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P373043 NM_013898.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138283 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167291 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14623760 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14623759 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14623761 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14525993 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14526001 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672454 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672453 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672451 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672455 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672412 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672447 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672437 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672448 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672440 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672426 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672444 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672439 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672442 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672425 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14641397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672438 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672436 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15140985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672457 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672423 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15051980 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10606691 - -
          Affymetrix Mouse Gene 1.0 ST Array 10361761 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606692 - -
          Affymetrix Mouse Gene 1.0 ST Array 10361762 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606690 - -
          Affymetrix HT MG-430 PM Array Plate 1416345_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1416346_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10606689 - -
          Affymetrix Mouse Gene 1.1 ST Array 10361760 - -
          Mouse Genome 430 2.0 Array 1416346_at - -
          Mouse Genome 430 2.0 Array 1416345_at - -
          Mouse Genome 430 2.0 Array 1416346_PM_at - -
          Mouse Genome 430 2.0 Array 1416345_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00018657.mm.1 TC0X00002993.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00018657.mm.1 NM_013898.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018656.mm.1 TC0X00002993.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00018656.mm.1 NM_013898.3 Refseq
          Mouse Expression Set 430 1416346_at - -
          Mouse Expression Set 430 1416345_at - -
          Mouse Expression Set 430 1416346_PM_at - -
          Mouse Expression Set 430 1416345_PM_at - -
          Murine Genome U74v2 Set 96849_PM_at - -
          Murine Genome U74v2 Set 96849_at - -
          Mouse Genome 430A 2.0 Array 1416345_PM_at - -
          Mouse Genome 430A 2.0 Array 1416346_at - -
          Mouse Genome 430A 2.0 Array 1416346_PM_at - -
          Mouse Genome 430A 2.0 Array 1416345_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 900209 - -
          ABI Mouse Genome Survey Microarray 900707 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 110279 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000110279 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2765922 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2765922 - -
          mouse-6 v1.1 (Illumina) scl53834.2_155-S - -
          Illumina Sentrix mouse-6.v1 bead array 110279 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53834.2_155-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 110279 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2896552 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2896552 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl53834.2_155-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b7b59cd5b-z8v7b:80/100.66.130.62:80.
          git-commit: 87deb889a5bb48f859ebd9970e87be562fd9396c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0