Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02603337_m1
          See other UGT1A1 GE Assays ›
          Gene Symbol:
          Ugt1a1
          Gene Name:
          UDP glucuronosyltransferase 1 family, polypeptide A1
          Gene Aliases:
          Gnt1, UDPGT 1-1, UGT1A01, Udpgt-1a, UgtBr1
          Chromosome Location:
          Chr.1: 88211959 - 88220002 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02603337_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ugt1a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_201645.2 NP_964007.2 1 - 2 914 135
          GenBank mRNA AA674349.1 - 1 - 2 159 135
          AY227194.1 - 1 - 2 914 135
          BC093516.1 - 1 - 2 876 5135493 135
          JX026002.1 - 1 - 2 871 135
          L02333.1 - 1 - 2 886 135
          S64760.1 - 1 - 2 886 135

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100000836.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ugt1a1

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          flavonoid biosynthetic process
          response to organic substance
          cellular response to hormone stimulus
          negative regulation of catalytic activity
          flavone metabolic process
          cellular glucuronidation
          flavonoid glucuronidation
          xenobiotic glucuronidation
          biphenyl catabolic process
          negative regulation of cellular glucuronidation
          retinoic acid binding
          enzyme inhibitor activity
          steroid binding
          glucuronosyltransferase activity
          transferase activity
          glycosyltransferase activity
          hexosyltransferase activity
          enzyme binding
          protein homodimerization activity
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P874080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422222 - -
          Whole Mouse Genome G4122A A_52_P874080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00516123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422222 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422240 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422236 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00114250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422188 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01132031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422205 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422233 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00270389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00516128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422224 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422215 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422227 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00715174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00270395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00516126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422221 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422200 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00270388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422234 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422217 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422225 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422247 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422248 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422199 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422195 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10348371 - -
          Affymetrix Mouse Gene 1.0 ST Array 10348375 - -
          Affymetrix Mouse Gene 1.0 ST Array 10348369 - -
          Affymetrix Mouse Gene 1.0 ST Array 10348372 - -
          Affymetrix Mouse Gene 1.0 ST Array 10348370 - -
          Affymetrix Mouse Gene 1.0 ST Array 10348373 - -
          Affymetrix Mouse Gene 1.0 ST Array 10348374 - -
          Affymetrix Mouse Gene 1.1 ST Array 10348354 - -
          Affymetrix Mouse Clariom S Array PSR0100007149.mm.1 TC0100000836.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100007149.mm.1 NM_201645.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100007149.mm.1 NM_201645.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100007148.mm.1 TC0100000836.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100007148.mm.1 NM_201645.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100007148.mm.1 NM_201645.2 Refseq
          Mouse Expression Set 430 1426261_s_at - -
          Mouse Expression Set 430 1426260_PM_a_at - -
          Mouse Expression Set 430 1424783_PM_a_at - -
          Mouse Expression Set 430 1426261_PM_s_at - -
          Mouse Expression Set 430 1426260_a_at - -
          Mouse Expression Set 430 1424783_a_at - -
          Murine Genome U74v2 Set 99580_PM_s_at - -
          Murine Genome U74v2 Set 99580_s_at - -
          Mouse Genome 430A 2.0 Array 1426260_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1426261_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1424783_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1426261_s_at - -
          Mouse Genome 430A 2.0 Array 1426260_a_at - -
          Mouse Genome 430A 2.0 Array 1424783_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 907248 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3140670 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228948 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228948 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
          git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline