Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02746061_sH
          See other HIST2H2BB GE Assays ›
          Gene Symbol:
          Hist2h2bb Hist1h2bg
          Gene Name:
          histone cluster 2, H2bb
          histone cluster 1, H2bg
          Gene Aliases:
          H2b-616
          -
          Chromosome Location:
          Chr.3: 96269700 - 96270192 on Build GRCm38
          Chromosome Location:
          Chr.13: 23571400 - 23571863 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm02746061_sH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hist2h2bb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175666.2 NP_783597.2 1 348 70
          Gene Symbol: Hist1h2bg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK134918.1 - 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000935.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hist2h2bb
          Click here to find siRNAs for Gene Hist1h2bg

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleosome assembly
          DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22348542 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22348539 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22348541 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22348534 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22348540 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2036883 NM_175666.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2036883 AK134918.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P361620 BC019122.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P361620 BC019122.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174497 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2036883 NM_175666.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2036883 AK134918.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P361620 BC019122.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P361620 BC019122.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22348535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22348542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22348544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22348539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22348541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22348537 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174497 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00704051 - -
          Whole Mouse Genome G4122A A_51_P361620 BC019122.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P361620 BC019122.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113833 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133426 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106973 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174497 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02953998 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02953995 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02953980 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02953999 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22348534 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22348541 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22348540 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22348539 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22348542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02607342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02953992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02953980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02810398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02810399 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03160164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02953974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03160167 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02953998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348542 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10494408 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494409 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494410 - -
          Affymetrix HT MG-430 PM Array Plate 1425868_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10494407 - -
          Mouse Genome 430 2.0 Array 1425868_PM_at - -
          Mouse Genome 430 2.0 Array 1425868_at - -
          Affymetrix Mouse Clariom S Array PSR0300007474.mm.1 TC0300000935.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300007474.mm.1 NM_175666.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007474.mm.1 NM_175666.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007475.mm.1 TC0300000935.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300007475.mm.1 NM_175666.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007475.mm.1 NM_175666.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007476.mm.1 TC0300000935.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300007476.mm.1 NM_175666.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007476.mm.1 NM_175666.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007477.mm.1 TC0300000935.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300007477.mm.1 NM_175666.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007477.mm.1 NM_175666.2 Refseq
          Mouse Expression Set 430 1425868_at - -
          Mouse Expression Set 430 1425868_PM_at - -
          Murine Genome U74v2 Set 94638_PM_at - -
          Murine Genome U74v2 Set 94638_at - -
          Mouse Genome 430A 2.0 Array 1425868_PM_at - -
          Mouse Genome 430A 2.0 Array 1425868_at - -
          Affymetrix HT Mouse Genome 430A Array 1425868_at - -
          Affymetrix HT Mouse Genome 430A Array 1425868_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 867713 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4210022 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004210022 - -
          mouse-6 v1.1 (Illumina) scl22867.1.51_23-S - -
          Illumina Sentrix mouse-6.v1 bead array 4210022 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl22867.1.51_23-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4210022 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1231066 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2817214 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1231066 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2817214 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl22867.1.51_23-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0