Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03015804_m1
          See other ZFP180 GE Assays ›
          Gene Symbol:
          Zfp180
          Gene Name:
          zinc finger protein 180
          Gene Aliases:
          2310040I01Rik, AI852378, D130011P11, HHZ168
          Chromosome Location:
          Chr.7: 24081897 - 24107709 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03015804_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp180
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001045486.2 NP_001038951.1 4 - 5 621 77
          NM_001289634.1 NP_001276563.1 5 - 6 725 77
          NM_001289637.1 NP_001276566.1 3 - 4 540 77
          NM_001289640.1 NP_001276569.1 4 - 5 616 77
          NM_001289641.1 NP_001276570.1 4 - 5 581 77
          GenBank mRNA AK141922.1 - 4 - 5 574 77
          AK144892.1 - 4 - 5 582 77
          AK145004.1 - 4 - 5 586 77
          AK154182.1 - 4 - 5 578 77
          AY166794.1 - 1 - 2 81 77
          BC063741.1 - 4 - 5 559 6504558 77
          BC066115.1 - 4 - 5 583 30619470 77
          BC085472.1 - 4 - 5 594 6417473 77
          CF736119.1 - 3 - 4 475 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000368.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170414 BC085472.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170414 BC066115.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170414 BC063741.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170414 AK154182.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170414 AK145004.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 677151 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 672041 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904009 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 743574 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 652791 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 36676 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 468680 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 618201 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 355705 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 632380 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 936212 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 106666 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 99536 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P180307 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136289 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170414 BC085472.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170414 BC066115.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170414 BC063741.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170414 AK154182.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170414 AK145004.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P157737 BC085472.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P157737 BC066115.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P157737 BC063741.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P157737 AK144892.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P157737 AK083802.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P224818 BC085472.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P224818 BC066115.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P224818 BC063741.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P224818 AK154182.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P224818 AY166794.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06103636 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06345404 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06345405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06144909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06329543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06329546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06380141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05990282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06329541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06329547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06314386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06460998 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10550836 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550834 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550838 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550839 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550837 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550835 - -
          Affymetrix HT MG-430 PM Array Plate 1428170_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10550833 - -
          Mouse Genome 430 2.0 Array 1428170_PM_at - -
          Mouse Genome 430 2.0 Array 1428170_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 210135_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 210135_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700002659.mm.1 TC0700000368.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002659.mm.1 NM_001045486.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002659.mm.1 NM_001289634.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002659.mm.1 NM_001289637.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002659.mm.1 NM_001289640.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002659.mm.1 NM_001289641.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002659.mm.1 NM_172483.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002660.mm.1 TC0700000368.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002660.mm.1 NM_001045486.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002660.mm.1 NM_001289634.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002660.mm.1 NM_001289637.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002660.mm.1 NM_001289640.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002660.mm.1 NM_001289641.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002660.mm.1 NM_172483.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002658.mm.1 TC0700000368.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002658.mm.1 NM_001045486.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002658.mm.1 NM_001289634.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002658.mm.1 NM_001289637.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002658.mm.1 NM_001289640.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002658.mm.1 NM_001289641.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002658.mm.1 NM_172483.2 Refseq
          Mouse Expression Set 430 1428170_PM_at - -
          Mouse Expression Set 430 1428170_at - -
          Murine Genome U74v2 Set 170098_PM_r_at - -
          Murine Genome U74v2 Set 170098_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1428170_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428170_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4150592 - -
          MouseWG-6 v2 Expression BeadChip 5720047 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2803368 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2803368 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5968f498c6-wvnc9:80/100.66.128.131:80.
          git-commit: b7f02e98b2654af9c52ff750aebd516a8b607fe3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/PDP_Cmrc_Card_Notification_Changes/CFGMAGJ/CFGMAGJ-5102/CFGMAGJ-5166-COMMERCE-CARD-TRANSLATION_APAC