Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03015865_g1
          See other RPL38 GE Assays ›
          Gene Symbol:
          Rpl38
          Gene Name:
          ribosomal protein L38
          Gene Aliases:
          0610025G13Rik, Rbt, Ts, Tss
          Chromosome Location:
          Chr.11: 114668543 - 114672331 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03015865_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl38
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001048057.1 NP_001041522.1 4 - 5 303 89
          NM_001048058.1 NP_001041523.1 3 - 4 229 89
          NM_023372.2 NP_075861.1 4 - 5 286 89
          GenBank mRNA AB037665.1 - 4 - 5 305 89
          AK002659.1 - 4 - 5 253 89
          AK010277.1 - 4 - 5 240 89
          AK010518.2 - 4 - 5 239 89
          AK012391.1 - 4 - 5 239 89
          AK151517.1 - 4 - 5 240 89
          BC034339.1 - 4 - 5 242 3985781 89
          BC047046.1 - 3 - 4 229 89
          BC055346.1 - 4 - 5 238 5003663 89
          BC081447.1 - 3 - 4 203 89
          BC094258.1 - 3 - 4 234 1245753 89
          BC094644.1 - 3 - 4 235 3328874 89
          BU921494.1 - 4 - 5 286 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001874.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpl38

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          ossification
          translation
          regulation of translation
          sensory perception of sound
          90S preribosome assembly
          middle ear morphogenesis
          axial mesoderm development
          molecular_function
          structural constituent of ribosome

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28149042 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28149046 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28149047 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28149048 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28149045 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2091308 BU921494.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2091308 BC094644.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2091308 NM_001048057.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2091308 NM_001048058.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2091308 NM_023372.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955059 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 875935 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 268135 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 23150 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P138024 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2091308 BU921494.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2091308 BC094644.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2091308 NM_001048057.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2091308 NM_001048058.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2091308 NM_023372.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955059 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P227332 BU921494.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P227332 BC094644.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P227332 NM_001048057.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P227332 NM_001048058.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P227332 NM_023372.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149042 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P138024 - -
          Whole Mouse Genome G4122A A_51_P227332 BU921494.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P227332 BC094644.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P227332 NM_001048057.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P227332 NM_001048058.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P227332 NM_023372.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127840 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P138024 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09428266 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09135146 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09428264 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28149046 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28149045 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28149047 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28149048 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28149042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28148999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09428264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09169985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09010217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09192025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28148996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09325190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09107181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09482218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09004208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149057 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10382370 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544813 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544814 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382373 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548977 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382375 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382372 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382371 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382374 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509561 - -
          Affymetrix Mouse Gene 1.1 ST Array 10509560 - -
          Affymetrix Mouse Gene 1.1 ST Array 10382369 - -
          Affymetrix Mouse Gene 1.1 ST Array 10548976 - -
          Affymetrix Mouse Gene 1.1 ST Array 10544812 - -
          Mouse Genome 430 2.0 Array 1433472_PM_x_at - -
          Mouse Genome 430 2.0 Array 1433472_x_at - -
          Affymetrix Mouse Clariom S Array PSR1100017516.mm.1 TC1100001874.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100017516.mm.1 NM_001048057.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017516.mm.1 NM_001048057.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017516.mm.1 NM_001048058.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017516.mm.1 NM_001048058.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017516.mm.1 NM_023372.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017516.mm.1 NM_023372.2 Refseq
          Mouse Expression Set 430 1433472_x_at - -
          Mouse Expression Set 430 1433472_PM_x_at - -
          Murine Genome U74v2 Set 99627_PM_r_at - -
          Murine Genome U74v2 Set 99626_PM_i_at - -
          Murine Genome U74v2 Set 99626_i_at - -
          Murine Genome U74v2 Set 99627_r_at - -
          Murine Genome U74v2 Set 169427_i_at - -
          Murine Genome U74v2 Set 169427_PM_i_at - -
          Mouse Genome 430A 2.0 Array 1433472_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1433472_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 898302 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4060138 - -
          MouseWG-6 v2 Expression BeadChip 70040 - -
          Illumina mouse-6 v1.0 expression beadchip 6200601 - -
          Illumina mouse-6 v1.0 expression beadchip 4060138 - -
          Illumina mouse-6 v1.0 expression beadchip 70040 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000070040 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004060138 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006200601 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2594971 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1223383 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223383 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2594971 - -
          mouse-6 v1.1 (Illumina) scl0067671.1_2-S - -
          mouse-6 v1.1 (Illumina) scl067671.4_8-S - -
          mouse-6 v1.1 (Illumina) scl40751.1_9-S - -
          Illumina Sentrix mouse-6.v1 bead array 6200601 - -
          Illumina Sentrix mouse-6.v1 bead array 70040 - -
          Illumina Sentrix mouse-6.v1 bead array 4060138 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0067671.1_2-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40751.1_9-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067671.4_8-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 70040 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6200601 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4060138 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2594971 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3135931 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2594971 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3135931 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl40751.1_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl067671.4_8-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0