Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03030145_gH
          See other IFNA1 GE Assays ›
          Gene Symbol:
          Ifna1 Ifna5 Ifna6
          Gene Name:
          interferon alpha 1
          interferon alpha 5
          interferon alpha 6
          Gene Aliases:
          Ifa1
          Ifa5
          IFNa8/6, Ifa6, Ifa8, Ifna8
          Chromosome Location:
          Chr.4: 88850087 - 88850656 on Build GRCm38
          Chromosome Location:
          Chr.4: 88835525 - 88836094 on Build GRCm38
          Chromosome Location:
          Chr.4: 88827416 - 88827985 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Mm03030145_gH
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ifna1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010502.2 NP_034632.2 1 42 74
          Gene Symbol: Ifna5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010505.2 NP_034635.2 1 42 74
          GenBank mRNA BC120910.1 - 1 42 40091152 74
          BC120911.1 - 1 42 40091153 74
          Gene Symbol: Ifna6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC151087.1 - 1 182 9088000 74

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ifna1
          Click here to find siRNAs for Gene Ifna5
          Click here to find siRNAs for Gene Ifna6

          Panther Classification:

          Molecular Function -

          interferon superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          T cell activation involved in immune response
          natural killer cell activation involved in immune response
          defense response
          humoral immune response
          cytokine-mediated signaling pathway
          B cell differentiation
          positive regulation of peptidyl-serine phosphorylation of STAT protein
          B cell proliferation
          defense response to bacterium
          response to exogenous dsRNA
          defense response to virus
          cytokine activity
          cytokine receptor binding
          type I interferon receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030174 NM_206867.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030174 BC114392.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030174 NM_010502.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030174 NM_206867.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030174 BC114392.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030174 NM_010502.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092512 - -
          Whole Mouse Genome G4122A A_51_P436401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03696015 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03898476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092544 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03845150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04078904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092571 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03433866 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03227261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03433868 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03417626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03748095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092526 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10514291 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514290 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505880 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505893 - -
          Affymetrix HT MG-430 PM Array Plate 1450564_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10505879 - -
          Affymetrix Mouse Gene 1.1 ST Array 10505892 - -
          Mouse Genome 430 2.0 Array 1450564_x_at - -
          Mouse Genome 430 2.0 Array 1450564_PM_x_at - -
          Affymetrix Mouse Clariom S Array PSR0400006102.mm.1 NM_206871.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006102.mm.1 NM_206871.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006102.mm.1 TC0400000906.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400006105.mm.1 NM_010505.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006105.mm.1 NM_010505.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006105.mm.1 TC0400000907.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400006111.mm.1 NM_010502.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006111.mm.1 NM_010502.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006111.mm.1 TC0400000909.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400006100.mm.1 TC0400000906.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400006104.mm.1 TC0400000906.mm.1 MTA TC
          Mouse Expression Set 430 1422404_x_at - -
          Mouse Expression Set 430 1450564_PM_x_at - -
          Mouse Expression Set 430 1450593_at - -
          Mouse Expression Set 430 1450564_x_at - -
          Mouse Expression Set 430 1450593_PM_at - -
          Mouse Expression Set 430 1422403_PM_at - -
          Mouse Expression Set 430 1422403_at - -
          Mouse Expression Set 430 1422404_PM_x_at - -
          Murine Genome U74v2 Set 102149_PM_f_at - -
          Murine Genome U74v2 Set 101619_at - -
          Murine Genome U74v2 Set 102149_f_at - -
          Murine Genome U74v2 Set 101788_f_at - -
          Murine Genome U74v2 Set 101619_PM_at - -
          Murine Genome U74v2 Set 101788_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1422404_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1450593_at - -
          Mouse Genome 430A 2.0 Array 1422404_x_at - -
          Mouse Genome 430A 2.0 Array 1450593_PM_at - -
          Mouse Genome 430A 2.0 Array 1450564_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1422403_at - -
          Mouse Genome 430A 2.0 Array 1422403_PM_at - -
          Mouse Genome 430A 2.0 Array 1450564_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1450564_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1450564_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 896817 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 100540592 - -
          Illumina mouse-6 v1.0 expression beadchip 5390592 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005390592 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1233604 - -
          mouse-6 v1.1 (Illumina) GI_6754289-S - -
          mouse-6 v1.1 (Illumina) scl015962.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 100540592 - -
          Illumina Sentrix mouse-6.v1 bead array 5390592 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl015962.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5390592 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100540592 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2858639 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1233604 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2858639 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2617487 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0