Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03031126_g1
          See other RNASEH2A GE Assays ›
          Gene Symbol:
          Rnaseh2a
          Gene Name:
          ribonuclease H2, large subunit
          Gene Aliases:
          2400006P09Rik, RNASEHI, RNHIA, RNHL
          Chromosome Location:
          Chr.8: 84956610 - 84969375 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03031126_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnaseh2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027187.3 NP_081463.1 3 - 4 356 78
          GenBank mRNA AK010292.1 - 3 - 4 346 78
          AK146518.1 - 3 - 4 357 78
          BC029252.1 - 3 - 4 386 3666709 78
          BC038158.1 - 3 - 4 430 1493985 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800002596.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnaseh2a

          Gene Ontology Categories:

          Function(s) Process(es)

          mismatch repair
          RNA catabolic process
          RNA metabolic process
          DNA replication, removal of RNA primer
          RNA phosphodiester bond hydrolysis, endonucleolytic
          nucleic acid binding
          RNA binding
          nuclease activity
          endonuclease activity
          RNA-DNA hybrid ribonuclease activity
          bidentate ribonuclease III activity
          hydrolase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022870 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022890 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022871 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022892 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022891 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022869 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966883 AK146518.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966883 AK010292.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966883 BC038158.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966883 NM_027187.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966878 AK146518.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966878 NM_027187.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115119 AK146518.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115119 AK220001.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115119 BC029252.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115119 AK010292.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115119 BC038158.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030749 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 396751 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323584 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 377016 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 41066 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 925791 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966883 AK146518.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966883 AK010292.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966883 BC038158.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966883 NM_027187.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115119 AK146518.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115119 AK220001.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115119 BC029252.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115119 AK010292.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115119 BC038158.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030749 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966878 AK146518.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966878 NM_027187.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022872 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106884 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06579699 - -
          Whole Mouse Genome G4122A A_51_P424128 BC029252.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424128 BC038158.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P676108 AK146518.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P676108 AK010292.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P676108 BC038158.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P676108 NM_027187.3 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106884 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06914269 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06914281 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07164740 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07128272 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07220107 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06922247 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07164715 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06914282 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06914280 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06922246 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022890 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022869 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022891 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022871 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022870 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07128272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07039397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07389546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06914273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07220107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07039396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06957660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07435644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06826037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022872 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10580278 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580280 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580279 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580277 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580275 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580276 - -
          Affymetrix HT MG-430 PM Array Plate 1452725_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10580274 - -
          Mouse Genome 430 2.0 Array 1452725_PM_a_at - -
          Mouse Genome 430 2.0 Array 1452725_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69724_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69724_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800019910.mm.1 TC0800002596.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800019910.mm.1 NM_027187.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019910.mm.1 NM_027187.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019918.mm.1 TC0800002596.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800019918.mm.1 NM_027187.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019918.mm.1 NM_027187.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019909.mm.1 TC0800002596.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800019909.mm.1 NM_027187.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019909.mm.1 NM_027187.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019911.mm.1 TC0800002596.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800019911.mm.1 NM_027187.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019911.mm.1 NM_027187.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019907.mm.1 TC0800002596.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800019907.mm.1 NM_027187.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019907.mm.1 NM_027187.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019913.mm.1 TC0800002596.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800019913.mm.1 NM_027187.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019913.mm.1 NM_027187.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019914.mm.1 TC0800002596.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800019914.mm.1 NM_027187.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019914.mm.1 NM_027187.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019916.mm.1 TC0800002596.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800019916.mm.1 NM_027187.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019916.mm.1 NM_027187.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019901.mm.1 TC0800002596.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800019901.mm.1 NM_027187.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019901.mm.1 NM_027187.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019920.mm.1 TC0800002596.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800019903.mm.1 TC0800002596.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800019903.mm.1 NM_027187.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019903.mm.1 NM_027187.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019915.mm.1 TC0800002596.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800019915.mm.1 NM_027187.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019915.mm.1 NM_027187.3 Refseq
          Mouse Expression Set 430 1452725_PM_a_at - -
          Mouse Expression Set 430 1452725_a_at - -
          Murine Genome U74v2 Set 96106_at - -
          Murine Genome U74v2 Set 96106_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1452725_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1452725_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4850270 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2746721 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2746721 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0