Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03053542_s1
          See other DHH GE Assays ›
          Gene Symbol:
          Dhh
          Gene Name:
          desert hedgehog
          Gene Aliases:
          C78960
          Chromosome Location:
          Chr.15: 98881630 - 98898572 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03053542_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dhh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007857.4 NP_031883.1 3 2300 126
          GenBank mRNA AK032466.1 - 3 2284 126
          AK033392.1 - 3 2302 126
          BE200181.1 - 1 126

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dhh

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          cell communication
          cell-cell signaling
          multicellular organism development
          spermatid development
          intein-mediated protein splicing
          male sex determination
          Leydig cell differentiation
          regulation of steroid biosynthetic process
          patched binding
          calcium ion binding
          protein binding
          peptidase activity
          zinc ion binding
          hydrolase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477182 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477212 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477210 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477178 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477185 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477179 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477211 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477213 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477186 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477181 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477187 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P232637 NM_007857.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P232637 AK032466.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P232637 AK033392.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P232637 NM_007857.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P232637 AK032466.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P232637 AK033392.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 676601 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169742 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 174904 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 792510 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 180589 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120412 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P232637 NM_007857.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P232637 AK032466.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P232637 AK033392.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P232637 NM_007857.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P232637 AK032466.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P232637 AK033392.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P232637 NM_007857.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P232637 AK032466.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P232637 AK033392.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P232637 NM_007857.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P232637 AK032466.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P232637 AK033392.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30477204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30477234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30477211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30477216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30477231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30477228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30477214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30477212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30477201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30477215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30477217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30477218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30477209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30477223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30477220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30477198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30477219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30477225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30477206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30477232 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08477510 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120412 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03112373 - -
          Whole Mouse Genome G4122A A_52_P232637 NM_007857.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P232637 AK032466.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P232637 AK033392.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P232637 NM_007857.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P232637 AK032466.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P232637 AK033392.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111542 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08477510 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120412 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11905472 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12302641 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12302643 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11905483 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477178 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477211 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477179 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477213 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477212 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477185 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477181 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477187 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477186 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477182 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30477179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477215 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11905480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12390248 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12044591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12346726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12126402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11905476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12126405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12126403 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12395592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12464997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11988317 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477231 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477190 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10432374 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432376 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432377 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432375 - -
          Affymetrix HT MG-430 PM Array Plate 1434959_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1422127_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10432372 - -
          Mouse Genome 430 2.0 Array 1422127_at - -
          Mouse Genome 430 2.0 Array 1434959_at - -
          Mouse Genome 430 2.0 Array 1434959_PM_at - -
          Mouse Genome 430 2.0 Array 1422127_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13363_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13363_at - -
          Mouse Expression Set 430 1422127_PM_at - -
          Mouse Expression Set 430 1422127_at - -
          Mouse Expression Set 430 1434959_PM_at - -
          Mouse Expression Set 430 1434959_at - -
          Murine Genome U74v2 Set 99861_at - -
          Murine Genome U74v2 Set 161588_r_at - -
          Murine Genome U74v2 Set 161111_PM_f_at - -
          Murine Genome U74v2 Set 161588_PM_r_at - -
          Murine Genome U74v2 Set 161111_f_at - -
          Murine Genome U74v2 Set 99861_PM_at - -
          Murine Genome U74v2 Set 134824_PM_s_at - -
          Murine Genome U74v2 Set 134825_r_at - -
          Murine Genome U74v2 Set 134825_PM_r_at - -
          Murine Genome U74v2 Set 134824_s_at - -
          Mouse Genome 430A 2.0 Array 1422127_at - -
          Mouse Genome 430A 2.0 Array 1434959_PM_at - -
          Mouse Genome 430A 2.0 Array 1422127_PM_at - -
          Mouse Genome 430A 2.0 Array 1434959_at - -
          Affymetrix HT Mouse Genome 430A Array 1422127_at - -
          Affymetrix HT Mouse Genome 430A Array 1434959_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422127_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1434959_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 507232 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2120433 - -
          Illumina mouse-6 v1.0 expression beadchip 101690044 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002120433 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2595664 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2595664 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2437411 - -
          mouse-6 v1.1 (Illumina) scl46746.3_409-S - -
          mouse-6 v1.1 (Illumina) scl0013363.2_203-S - -
          Illumina Sentrix mouse-6.v1 bead array 101690044 - -
          Illumina Sentrix mouse-6.v1 bead array 2120433 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0013363.2_203-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2120433 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101690044 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2595664 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2595664 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0