Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03455591_m1
          See other IGF2OS GE Assays ›
          Gene Symbol:
          Igf2os
          Gene Name:
          insulin-like growth factor 2, opposite strand
          Gene Aliases:
          Igf2as, Peg8
          Chromosome Location:
          Chr.7: 142659693 - 142670356 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03455591_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Igf2os
          Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002855.2
          3 - 4 1614 78
          GenBank mRNA AB030734.1 - LIT1673
          PEG8/Igf2as
          3 - 4 1613 78

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream insulin-like growth factor 2 Igf2 AL033362,
          Igf-2,
          Igf-II,
          M6pr,
          Mpr,
          Peg2
          -4.0kb Anti-sense Chr. 7 - 142650768 - 142662270 on NCBI Build GRCm38
          Downstream insulin II Ins2 AA986540,
          Ins-2,
          InsII,
          Mody,
          Mody4,
          proinsulin
          8.3kb Anti-sense Chr. 7 - 142678656 - 142679708 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002084.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P116635 NR_002855.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P116635 AB030734.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 30515 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 86007 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 578801 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 520319 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 649068 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 79852 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P116635 NR_002855.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P116635 AB030734.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06399927 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01657950 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06399913 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01657944 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130311 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116635 NR_002855.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116635 AB030734.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01657950 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10559240 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559242 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559239 - -
          Affymetrix HT MG-430 PM Array Plate 1427394_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10559241 - -
          Affymetrix Mouse Gene 1.1 ST Array 10559238 - -
          Mouse Genome 430 2.0 Array 1427394_at - -
          Mouse Genome 430 2.0 Array 1427394_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 111975_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 111975_at - -
          Affymetrix Mouse Clariom S Array PSR0700018777.mm.1 TC0700002084.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700018777.mm.1 NR_002855.3 Refseq
          Mouse Expression Set 430 1427394_at - -
          Mouse Expression Set 430 1427394_PM_at - -
          Mouse Genome 430A 2.0 Array 1427394_PM_at - -
          Mouse Genome 430A 2.0 Array 1427394_at - -
          Affymetrix HT Mouse Genome 430A Array 1427394_at - -
          Affymetrix HT Mouse Genome 430A Array 1427394_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 476499 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 107050170 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1235307 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0