Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03455899_m1
          See other C230091D08RIK GE Assays ›
          Gene Symbol:
          C230091D08Rik
          Gene Name:
          RIKEN cDNA C230091D08 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 59307924 - 59324149 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03455899_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C230091D08Rik
          Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015479.1
          2 - 3 1893 139
          GenBank mRNA AK049008.1 - LIT3431
          UBE3A-ATS
          2 - 3 1893 139

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ubiquitin protein ligase E3A Ube3a 4732496B02,
          5830462N02Rik,
          A130086L21Rik,
          Hpve6a,
          mKIAA4216
          Anti-sense Chr. 7 - 59227814 - 59311536 on NCBI Build GRCm38
          Downstream imprinted gene in the Prader-Willi syndrome region Ipw 301.1kb Sense Chr. 7 - 59619158 - 59678878 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003126.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141813 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162289 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06211158 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162289 - -
          Whole Mouse Genome G4122A A_52_P584458 AK140886.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162289 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1438695_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1437354_PM_at - -
          Mouse Genome 430 2.0 Array 1438695_PM_at - -
          Mouse Genome 430 2.0 Array 1437354_PM_at - -
          Mouse Genome 430 2.0 Array 1438695_at - -
          Mouse Genome 430 2.0 Array 1437354_at - -
          Affymetrix Mouse Clariom S Array PSR0700028265.mm.1 TC0700003126.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700028265.mm.1 NR_015479.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700028265.mm.1 NR_015479.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700028264.mm.1 TC0700003126.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700028264.mm.1 NR_015479.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700028264.mm.1 NR_015479.1 Refseq
          Mouse Expression Set 430 1438695_PM_at - -
          Mouse Expression Set 430 1438695_at - -
          Mouse Expression Set 430 1437354_at - -
          Mouse Expression Set 430 1437354_PM_at - -
          Mouse Genome 430A 2.0 Array 1437354_PM_at - -
          Mouse Genome 430A 2.0 Array 1438695_at - -
          Mouse Genome 430A 2.0 Array 1437354_at - -
          Mouse Genome 430A 2.0 Array 1438695_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1438695_at - -
          Affymetrix HT Mouse Genome 430A Array 1438695_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1437354_at - -
          Affymetrix HT Mouse Genome 430A Array 1437354_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b7b59cd5b-z8v7b:80/100.66.130.62:80.
          git-commit: 87deb889a5bb48f859ebd9970e87be562fd9396c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0