Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03456298_g1
          See other MEG3 GE Assays ›
          Gene Symbol:
          Meg3
          Gene Name:
          maternally expressed 3
          Gene Aliases:
          2900016C05Rik, 3110050O07Rik, 6330408G06Rik, AI425946, AW108224, D12Bwg1266e, Gtl2, R74756, R75394
          Chromosome Location:
          Chr.12: 109540996 - 109571729 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03456298_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Meg3
          Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027652.1
          6 - 7 1256 62
          GenBank mRNA BC051667.1 -
          420 62
          Y13832.1 - LIT3453
          Gtl2
          6 - 7 1247 62
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CB194103.1 - 1 - 2 81 Chr. 12 - 109549289 - 109571721 [+] on Build GRCm38 62
          GenBankmRNA CB183928.1 - 706 Chr. - - [-] on Build 62
          GenBankmRNA CF584712.1 - 460 Chr. - - [-] on Build 62
          GenBankmRNA BP775209.1 - 2 - 3 Chr. 12 - 109544722 - 109549805 [-] on Build GRCm38 62
          GenBankmRNA AA183389.1 - 1 - 2 101 Chr. 12 - 109549269 - 109558284 [+] on Build GRCm38 62
          GenBankmRNA BM053819.1 - 2 - 3 266 Chr. 12 - 109546356 - 109549803 [+] on Build GRCm38 62
          GenBankmRNA AV469420.1 - 306 Chr. - - [-] on Build 62
          GenBankmRNA BU058056.1 - 405 Chr. - - [-] on Build 62
          GenBankmRNA CK030896.1 - 3 - 4 361 Chr. 12 - 109544574 - 109571455 [+] on Build GRCm38 62
          GenBankmRNA DN174347.1 - 3 - 4 273 Chr. 12 - 109544662 - 109568602 [+] on Build GRCm38 62
          GenBankmRNA CB202480.1 - 299 Chr. - - [-] on Build 62
          GenBankmRNA BI441350.1 - 2 - 3 294 Chr. 12 - 109546328 - 109549691 [+] on Build GRCm38 62
          GenBankmRNA BF658505.1 - 1 - 2 115 Chr. 12 - 109549255 - 109571463 [+] on Build GRCm38 62
          GenBankmRNA AV583825.1 - 3 - 4 247 Chr. 12 - 109544985 - 109558255 [+] on Build GRCm38 62
          GenBankmRNA AV584558.1 - 1 - 2 136 Chr. 12 - 109549234 - 109571423 [+] on Build GRCm38 62
          GenBankmRNA CF578092.1 - 4 - 5 Chr. 12 - 109549171 - 109571726 [-] on Build GRCm38 62
          GenBankmRNA CB059150.1 - 1 - 2 150 Chr. 12 - 109549228 - 109571595 [+] on Build GRCm38 62
          GenBankmRNA CF584777.1 - 441 Chr. - - [-] on Build 62
          GenBankmRNA CF585122.1 - 2 - 3 322 Chr. 12 - 109546306 - 109571648 [+] on Build GRCm38 62
          GenBankmRNA AA033444.1 - 1 - 2 100 Chr. 12 - 109549270 - 109558295 [+] on Build GRCm38 62
          GenBankmRNA CB058206.1 - 3 - 4 439 Chr. 12 - 109544498 - 109549697 [+] on Build GRCm38 62
          GenBankmRNA BI965904.1 - 2 - 3 266 Chr. 12 - 109546357 - 109549793 [+] on Build GRCm38 62
          GenBankmRNA AV480686.1 - 1 - 2 156 Chr. 12 - 109549224 - 109558320 [+] on Build GRCm38 62
          GenBankmRNA CK019912.1 - 411 Chr. - - [-] on Build 62
          GenBankmRNA BM894630.1 - 1 - 2 82 Chr. 12 - 109549288 - 109571555 [+] on Build GRCm38 62
          GenBankmRNA BM053951.1 - 2 - 3 266 Chr. 12 - 109546356 - 109549790 [+] on Build GRCm38 62
          GenBankmRNA AV502014.1 - 1 - 2 90 Chr. 12 - 109549280 - 109558307 [+] on Build GRCm38 62
          GenBankmRNA BE855234.1 - 1 - 2 181 Chr. 12 - 109549189 - 109558320 [+] on Build GRCm38 62
          GenBankmRNA CF618073.1 - 1 - 2 237 Chr. 12 - 109549157 - 109571607 [+] on Build GRCm38 62
          GenBankmRNA BI965842.1 - 2 - 3 282 Chr. 12 - 109546340 - 109549809 [+] on Build GRCm38 62

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 1906-1 Mir1906-1 Mir1906,
          Mirn1906,
          mmu-mir-1906,
          mmu-mir-1906-1
          -4.7kb Sense Chr. 12 - 109544541 - 109544620 on NCBI Build GRCm38
          Downstream microRNA 770 Mir770 Mirn770,
          mir-770,
          mmu-mir-770
          14.1kb Sense Chr. 12 - 109563692 - 109563785 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s)

          in utero embryonic development
          liver development
          DNA methylation
          skeletal muscle tissue development
          determination of adult lifespan
          post-embryonic development
          multicellular organism growth
          lung alveolus development
          SMAD protein signal transduction
          genetic imprinting

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0