Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03646584_g1
          See other RD3L GE Assays ›
          Gene Symbol:
          Rd3l
          Gene Name:
          retinal degeneration 3-like
          Gene Aliases:
          A930011N13
          Chromosome Location:
          Chr.12: 111979191 - 111981128 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03646584_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rd3l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001127685.1 NP_001121157.1 2 - 3 335 123
          GenBank mRNA BC048943.1 - 3 - 4 591 123
          BC147034.1 - 2 - 3 335 9056031 123
          BC147035.1 - 2 - 3 335 9056043 123
          BC147747.1 - 2 - 3 335 9056696 123
          BC147749.1 - 2 - 3 335 9056698 123

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200002422.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2110250 BC147035.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2110250 BC147747.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2110250 BC147749.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2110250 NM_001127685.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2110250 AK080710.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2110250 BC147035.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2110250 BC147747.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2110250 BC147749.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2110250 NM_001127685.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2110250 AK080710.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747235 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09964221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747269 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747272 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10034441 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09964228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09964220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747263 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747256 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747275 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747274 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747226 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09820136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09964219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747271 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09802916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747231 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747240 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200016829.mm.1 TC1200002422.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200016829.mm.1 NM_001127685.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016829.mm.1 NM_001127685.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016828.mm.1 TC1200002422.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200016828.mm.1 NM_001127685.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016828.mm.1 NM_001127685.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016826.mm.1 TC1200002422.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200016826.mm.1 NM_001127685.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016826.mm.1 NM_001127685.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 446290 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1980133 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2673745 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2673745 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0