Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03650010_s1
          See other GM14057 GE Assays ›
          Gene Symbol:
          Gm14057
          Gene Name:
          Ppp1r14c pseudogene
          Gene Aliases:
          OTTMUSG00000015563
          Chromosome Location:
          Chr.2: 130801966 - 130804101 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03650010_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm14057
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_024097.1 1 150 121
          GenBank mRNA BC057040.1 - 1737 121

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream solute carrier family 4,
          sodium bicarbonate transporter-like,
          member 11
          Slc4a11 AI503023,
          BTR1,
          NaBC1
          -104.4kb Sense Chr. 2 - 130684107 - 130697519 on NCBI Build GRCm38
          Downstream RIKEN cDNA A730017L22 gene A730017L22Rik 68.4kb Sense Chr. 2 - 130872541 - 130906396 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200005505.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139172 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1430285_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1453465_PM_x_at - -
          Mouse Genome 430 2.0 Array 1430285_PM_at - -
          Mouse Genome 430 2.0 Array 1430286_PM_s_at - -
          Mouse Genome 430 2.0 Array 1430285_at - -
          Mouse Genome 430 2.0 Array 1430286_s_at - -
          Mouse Genome 430 2.0 Array 1453465_PM_x_at - -
          Mouse Genome 430 2.0 Array 1453465_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 100043766_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 100043766_at - -
          Affymetrix Mouse Clariom S Array PSR0200039755.mm.1 TC0200005505.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039755.mm.1 NR_024097.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039757.mm.1 TC0200005505.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039757.mm.1 NR_024097.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039756.mm.1 TC0200005505.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039756.mm.1 NR_024097.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039754.mm.1 TC0200005505.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039754.mm.1 NR_024097.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039758.mm.1 TC0200005505.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039758.mm.1 NR_024097.1 Refseq
          Mouse Expression Set 430 1430285_PM_at - -
          Mouse Expression Set 430 1453465_PM_x_at - -
          Mouse Expression Set 430 1430285_at - -
          Mouse Expression Set 430 1430286_s_at - -
          Mouse Expression Set 430 1453465_x_at - -
          Mouse Expression Set 430 1430286_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1430285_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453465_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1453465_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1430285_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 893813 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0