Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03682027_s1
          See other 1700096K18RIK GE Assays ›
          Gene Symbol:
          1700096K18Rik
          Gene Name:
          RIKEN cDNA 1700096K18 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 25530018 - 25531466 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03682027_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700096K18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027388.1 1 451 108
          GenBank mRNA AK047471.1 - 1 452 108
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BE634397.1 - 1 75 Chr. 5 - 25530394 - 25530758 [+] on Build GRCm38 108
          GenBankmRNA CN831667.1 - 1 225 Chr. 5 - 25530247 - 25530770 [+] on Build GRCm38 108
          GenBankmRNA CN838866.1 - 1 454 Chr. 5 - 25530018 - 25530636 [+] on Build GRCm38 108
          GenBankmRNA CK623483.1 - 1 236 Chr. 5 - 25530233 - 25530766 [+] on Build GRCm38 108
          GenBankmRNA BB647168.1 - 1 448 Chr. 5 - 25530021 - 25530652 [+] on Build GRCm38 108
          GenBankmRNA BU939214.1 - 1 63 Chr. 5 - 25530408 - 25530770 [+] on Build GRCm38 108

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream chaperonin containing TCP1,
          subunit 8 (theta)-like 1
          Cct8l1 Gm443 -12.0kb Sense Chr. 5 - 25516067 - 25518027 on NCBI Build GRCm38
          Downstream X-ray repair complementing defective repair in Chinese hamster cells 2 Xrcc2 4921524O04Rik,
          8030409M04Rik,
          RAD51,
          RecA
          158.3kb Anti-sense Chr. 5 - 25689814 - 25705797 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000236.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2334112 NR_027388.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2334112 AK047471.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2334112 BE980748.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 23110 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2334112 NR_027388.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2334112 AK047471.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2334112 BE980748.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P448805 NR_027388.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P448805 AK047471.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P04806649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23547914 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23547916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04271805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23547926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23547937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23547929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04183787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23547932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23547915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04327238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23547942 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04183786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04271811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23547940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23547918 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1440953_PM_at - -
          Mouse Genome 430 2.0 Array 1440953_at - -
          Mouse Genome 430 2.0 Array 1440953_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500001830.mm.1 TC0500000236.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500001830.mm.1 NR_027388.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500001830.mm.1 NR_027388.1 Refseq
          Mouse Expression Set 430 1440953_at - -
          Mouse Expression Set 430 1440953_PM_at - -
          Murine Genome U74v2 Set 167570_at - -
          Murine Genome U74v2 Set 167570_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440953_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440953_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0